Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8006985-8007224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCY2D
Entrez Gene ID
3000
Gene Name
guanylate cyclase 2D, retinal
Gene Aliases
CORD5, CORD6, CYGD, GUC1A4, GUC2D, LCA, LCA1, RCD2, RETGC-1, ROS-GC1, ROSGC, retGC
UniGene
Hs.592109
Gene Chromosome Location
Chr.17: 8002670-8020340 on Build GRCh38
Transcripts
NM_000180.3 XM_011523816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
985793 c.1402C>A p.L468I Chr.17: 8007083-8007083 on Build GRCh38
3523508 c.1459G>A p.V487M Chr.17: 8007140-8007140 on Build GRCh38
355079 c.1390G>T p.G464C Chr.17: 8007071-8007071 on Build GRCh38
6148428 c.1410C>T p.F470F Chr.17: 8007091-8007091 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374138416 Chr.17: 8007025-8007025 on Build GRCh38
rs998623870 Chr.17: 8007118-8007118 on Build GRCh38
rs61749681 Chr.17: 8007112-8007113 on Build GRCh38
rs766944930 Chr.17: 8007114-8007114 on Build GRCh38
rs373533477 Chr.17: 8007040-8007040 on Build GRCh38
rs767033042 Chr.17: 8007019-8007019 on Build GRCh38
rs745618018 Chr.17: 8007076-8007076 on Build GRCh38
rs753166496 Chr.17: 8007135-8007135 on Build GRCh38
rs756787049 Chr.17: 8007161-8007161 on Build GRCh38
rs777136217 Chr.17: 8007039-8007039 on Build GRCh38
rs960653636 Chr.17: 8007107-8007107 on Build GRCh38
rs757158931 Chr.17: 8007052-8007052 on Build GRCh38
rs371908939 Chr.17: 8007024-8007024 on Build GRCh38
rs780858451 Chr.17: 8007028-8007028 on Build GRCh38
rs749966010 Chr.17: 8007051-8007052 on Build GRCh38
rs748301943 Chr.17: 8007086-8007086 on Build GRCh38
rs770119513 Chr.17: 8007098-8007098 on Build GRCh38
rs867041083 Chr.17: 8007104-8007104 on Build GRCh38
rs138298187 Chr.17: 8007113-8007113 on Build GRCh38
rs1005796154 Chr.17: 8007174-8007174 on Build GRCh38
rs141346556 Chr.17: 8007130-8007130 on Build GRCh38
rs771764405 Chr.17: 8007077-8007077 on Build GRCh38
rs190851646 Chr.17: 8007034-8007034 on Build GRCh38
rs767788419 Chr.17: 8007132-8007132 on Build GRCh38
rs763851107 Chr.17: 8007151-8007151 on Build GRCh38
rs771288323 Chr.17: 8007138-8007138 on Build GRCh38
rs745816219 Chr.17: 8007178-8007178 on Build GRCh38
rs753328828 Chr.17: 8007159-8007159 on Build GRCh38
rs371533971 Chr.17: 8007164-8007164 on Build GRCh38
rs773394059 Chr.17: 8007110-8007110 on Build GRCh38
rs61749680 Chr.17: 8007032-8007032 on Build GRCh38
rs112984002 Chr.17: 8007183-8007183 on Build GRCh38
rs779707457 Chr.17: 8007184-8007184 on Build GRCh38
rs1048867421 Chr.17: 8007176-8007176 on Build GRCh38
rs145060888 Chr.17: 8007137-8007137 on Build GRCh38
rs901228620 Chr.17: 8007106-8007106 on Build GRCh38
rs778875311 Chr.17: 8007055-8007055 on Build GRCh38
rs538719629 Chr.17: 8007041-8007041 on Build GRCh38
rs980774971 Chr.17: 8007017-8007017 on Build GRCh38
rs553866555 Chr.17: 8007044-8007044 on Build GRCh38
rs943098635 Chr.17: 8007179-8007179 on Build GRCh38
rs572166519 Chr.17: 8007127-8007127 on Build GRCh38

View Full Product Details