Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 176864085-176864346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
UniGene
Hs.495897
Gene Chromosome Location
Chr.1: 176855118-177164904 on Build GRCh38
Transcripts
NM_001286164.1 NM_004319.2 XM_017001340.1 XM_017001341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4874783 c.3896G>T p.S1299I Chr.1: 176864297-176864297 on Build GRCh38
3478943 c.3867G>A p.G1289G Chr.1: 176864302-176864302 on Build GRCh38
3478944 c.3849C>T p.I1283I Chr.1: 176864320-176864320 on Build GRCh38
3802960 c.3859G>A p.D1287N Chr.1: 176864310-176864310 on Build GRCh38
6527909 c.3876A>G p.K1292K Chr.1: 176864293-176864293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998911704 Chr.1: 176864206-176864206 on Build GRCh38
rs144312834 Chr.1: 176864135-176864135 on Build GRCh38
rs773194016 Chr.1: 176864294-176864294 on Build GRCh38
rs957399841 Chr.1: 176864129-176864129 on Build GRCh38
rs1051447887 Chr.1: 176864191-176864191 on Build GRCh38
rs950513828 Chr.1: 176864205-176864205 on Build GRCh38
rs965134639 Chr.1: 176864165-176864165 on Build GRCh38
rs1017689388 Chr.1: 176864164-176864164 on Build GRCh38
rs546011684 Chr.1: 176864124-176864124 on Build GRCh38
rs758538695 Chr.1: 176864273-176864273 on Build GRCh38
rs923128967 Chr.1: 176864187-176864187 on Build GRCh38
rs766576098 Chr.1: 176864311-176864311 on Build GRCh38
rs75385838 Chr.1: 176864196-176864196 on Build GRCh38
rs771250301 Chr.1: 176864267-176864268 on Build GRCh38
rs975955432 Chr.1: 176864185-176864185 on Build GRCh38
rs769290089 Chr.1: 176864270-176864270 on Build GRCh38
rs774677567 Chr.1: 176864299-176864310 on Build GRCh38
rs528148259 Chr.1: 176864151-176864151 on Build GRCh38
rs200991545 Chr.1: 176864279-176864279 on Build GRCh38
rs902870665 Chr.1: 176864195-176864195 on Build GRCh38
rs971531077 Chr.1: 176864296-176864296 on Build GRCh38
rs879675526 Chr.1: 176864266-176864266 on Build GRCh38
rs146353059 Chr.1: 176864319-176864319 on Build GRCh38
rs200271835 Chr.1: 176864318-176864318 on Build GRCh38
rs561493560 Chr.1: 176864234-176864234 on Build GRCh38
rs61756323 Chr.1: 176864291-176864291 on Build GRCh38
rs533386001 Chr.1: 176864321-176864321 on Build GRCh38
rs774165665 Chr.1: 176864289-176864289 on Build GRCh38
rs760721420 Chr.1: 176864310-176864310 on Build GRCh38
rs753049744 Chr.1: 176864320-176864320 on Build GRCh38
rs748957256 Chr.1: 176864275-176864275 on Build GRCh38
rs745410803 Chr.1: 176864256-176864256 on Build GRCh38
rs541870703 Chr.1: 176864210-176864210 on Build GRCh38

View Full Product Details