Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81512036-81512273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTG1
Entrez Gene ID
71
Gene Name
actin gamma 1
Gene Aliases
ACT, ACTG, BRWS2, DFNA20, DFNA26, HEL-176
UniGene
Hs.514581
Gene Chromosome Location
Chr.17: 81509971-81512866 on Build GRCh38
Transcripts
NM_001199954.1 NM_001614.3 NR_037688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4139949 c.136G>A p.G46S Chr.17: 81512130-81512130 on Build GRCh38
6048728 c.209_210insC p.I71fs*2 Chr.17: 81512056-81512057 on Build GRCh38
6082552 c.98C>T p.S33F Chr.17: 81512257-81512257 on Build GRCh38
4435434 c.102C>T p.I34I Chr.17: 81512253-81512253 on Build GRCh38
6082553 c.136G>T p.G46C Chr.17: 81512130-81512130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11549193 Chr.17: 81512250-81512250 on Build GRCh38
rs782763504 Chr.17: 81512214-81512214 on Build GRCh38
rs781879811 Chr.17: 81512190-81512190 on Build GRCh38
rs202223445 Chr.17: 81512207-81512207 on Build GRCh38
rs782796849 Chr.17: 81512219-81512219 on Build GRCh38
rs782275620 Chr.17: 81512244-81512244 on Build GRCh38
rs201275526 Chr.17: 81512056-81512056 on Build GRCh38
rs201951886 Chr.17: 81512172-81512172 on Build GRCh38
rs782512698 Chr.17: 81512233-81512233 on Build GRCh38
rs373618606 Chr.17: 81512148-81512148 on Build GRCh38
rs374907377 Chr.17: 81512128-81512128 on Build GRCh38
rs368299537 Chr.17: 81512098-81512098 on Build GRCh38
rs562047291 Chr.17: 81512241-81512241 on Build GRCh38
rs11549224 Chr.17: 81512067-81512067 on Build GRCh38
rs536476533 Chr.17: 81512224-81512224 on Build GRCh38
rs770592789 Chr.17: 81512189-81512189 on Build GRCh38
rs782100140 Chr.17: 81512211-81512211 on Build GRCh38
rs781886356 Chr.17: 81512181-81512182 on Build GRCh38
rs782754511 Chr.17: 81512065-81512065 on Build GRCh38
rs571472114 Chr.17: 81512174-81512174 on Build GRCh38
rs550758256 Chr.17: 81512203-81512203 on Build GRCh38
rs374327916 Chr.17: 81512159-81512159 on Build GRCh38
rs115307446 Chr.17: 81512156-81512156 on Build GRCh38
rs372586603 Chr.17: 81512095-81512095 on Build GRCh38
rs11549244 Chr.17: 81512242-81512242 on Build GRCh38
rs782351704 Chr.17: 81512201-81512201 on Build GRCh38
rs781910131 Chr.17: 81512171-81512171 on Build GRCh38
rs11549174 Chr.17: 81512235-81512235 on Build GRCh38
rs148759022 Chr.17: 81512087-81512087 on Build GRCh38
rs11549216 Chr.17: 81512105-81512105 on Build GRCh38
rs11549220 Chr.17: 81512089-81512089 on Build GRCh38
rs781861355 Chr.17: 81512216-81512216 on Build GRCh38
rs376850595 Chr.17: 81512151-81512151 on Build GRCh38
rs782660034 Chr.17: 81512149-81512149 on Build GRCh38
rs782654999 Chr.17: 81512084-81512084 on Build GRCh38
rs577515884 Chr.17: 81512117-81512117 on Build GRCh38
rs144338558 Chr.17: 81512086-81512086 on Build GRCh38
rs369910719 Chr.17: 81512195-81512195 on Build GRCh38
rs782205858 Chr.17: 81512092-81512092 on Build GRCh38
rs367960707 Chr.17: 81512209-81512209 on Build GRCh38
rs11549214 Chr.17: 81512124-81512124 on Build GRCh38
rs147032031 Chr.17: 81512068-81512068 on Build GRCh38
rs782566714 Chr.17: 81512141-81512141 on Build GRCh38
rs200210656 Chr.17: 81512129-81512129 on Build GRCh38
rs11549199 Chr.17: 81512090-81512090 on Build GRCh38
rs782713618 Chr.17: 81512110-81512110 on Build GRCh38
rs11549197 Chr.17: 81512140-81512140 on Build GRCh38
rs11549233 Chr.17: 81512064-81512064 on Build GRCh38
rs11549236 Chr.17: 81512236-81512236 on Build GRCh38
rs886041280 Chr.17: 81512135-81512135 on Build GRCh38
rs536366580 Chr.17: 81512062-81512062 on Build GRCh38
rs140152735 Chr.17: 81512076-81512076 on Build GRCh38
rs782815041 Chr.17: 81512113-81512113 on Build GRCh38
rs782608331 Chr.17: 81512147-81512147 on Build GRCh38
rs146402466 Chr.17: 81512101-81512101 on Build GRCh38
rs782798449 Chr.17: 81512168-81512168 on Build GRCh38
rs782119646 Chr.17: 81512253-81512253 on Build GRCh38
rs782370155 Chr.17: 81512158-81512158 on Build GRCh38
rs11549245 Chr.17: 81512077-81512077 on Build GRCh38
rs782504245 Chr.17: 81512082-81512082 on Build GRCh38
rs782129674 Chr.17: 81512166-81512166 on Build GRCh38
rs183867515 Chr.17: 81512206-81512206 on Build GRCh38
rs138240892 Chr.17: 81512074-81512074 on Build GRCh38
rs782127437 Chr.17: 81512200-81512200 on Build GRCh38
rs782481421 Chr.17: 81512146-81512146 on Build GRCh38
rs370205158 Chr.17: 81512202-81512202 on Build GRCh38
rs782606686 Chr.17: 81512227-81512227 on Build GRCh38
rs139751304 Chr.17: 81512107-81512107 on Build GRCh38
rs1057518673 Chr.17: 81512237-81512237 on Build GRCh38
rs201279208 Chr.17: 81512150-81512150 on Build GRCh38
rs782205195 Chr.17: 81512196-81512196 on Build GRCh38
rs782473240 Chr.17: 81512075-81512075 on Build GRCh38
rs781953399 Chr.17: 81512060-81512060 on Build GRCh38
rs782777443 Chr.17: 81512122-81512122 on Build GRCh38
rs782730692 Chr.17: 81512173-81512173 on Build GRCh38
rs143798951 Chr.17: 81512145-81512145 on Build GRCh38
rs782137311 Chr.17: 81512217-81512217 on Build GRCh38
rs377741704 Chr.17: 81512180-81512180 on Build GRCh38
rs782056092 Chr.17: 81512109-81512109 on Build GRCh38

View Full Product Details