Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 19517972-19518213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEBP2
Entrez Gene ID
121536
Gene Name
AE binding protein 2
Gene Aliases
-
UniGene
Hs.126497
Gene Chromosome Location
Chr.12: 19404101-19522239 on Build GRCh38
Transcripts
NM_001114176.1 NM_001267043.1 NM_153207.4 XM_006719031.3 XM_017018806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6595686 c.860+7A>C p.? Chr.12: 19518120-19518120 on Build GRCh38
6595685 c.1508+7A>C p.? Chr.12: 19518120-19518120 on Build GRCh38
2004311 c.1491G>A p.P497P Chr.12: 19518096-19518096 on Build GRCh38
2004313 c.1508+3delA p.? Chr.12: 19518116-19518116 on Build GRCh38
3935865 c.1484C>G p.T495R Chr.12: 19518089-19518089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752539638 Chr.12: 19518115-19518116 on Build GRCh38
rs547590825 Chr.12: 19518049-19518049 on Build GRCh38
rs749301707 Chr.12: 19518161-19518161 on Build GRCh38
rs187522462 Chr.12: 19518096-19518096 on Build GRCh38
rs936119968 Chr.12: 19518119-19518122 on Build GRCh38
rs535861276 Chr.12: 19518184-19518184 on Build GRCh38
rs773713644 Chr.12: 19518179-19518179 on Build GRCh38
rs757081762 Chr.12: 19518077-19518077 on Build GRCh38
rs550295518 Chr.12: 19518147-19518147 on Build GRCh38
rs371899888 Chr.12: 19518149-19518149 on Build GRCh38
rs973907630 Chr.12: 19518151-19518151 on Build GRCh38
rs757983455 Chr.12: 19518064-19518064 on Build GRCh38
rs183192816 Chr.12: 19518022-19518022 on Build GRCh38
rs570118272 Chr.12: 19518154-19518154 on Build GRCh38
rs533509061 Chr.12: 19518133-19518133 on Build GRCh38
rs374299102 Chr.12: 19518035-19518035 on Build GRCh38
rs762196820 Chr.12: 19518007-19518007 on Build GRCh38
rs950277159 Chr.12: 19518015-19518015 on Build GRCh38
rs760434888 Chr.12: 19518130-19518131 on Build GRCh38
rs929717888 Chr.12: 19518173-19518173 on Build GRCh38
rs1054971858 Chr.12: 19518150-19518150 on Build GRCh38
rs200916179 Chr.12: 19518037-19518037 on Build GRCh38
rs1046775523 Chr.12: 19518177-19518177 on Build GRCh38
rs752316059 Chr.12: 19518053-19518053 on Build GRCh38
rs12821385 Chr.12: 19518010-19518010 on Build GRCh38
rs781085334 Chr.12: 19518114-19518114 on Build GRCh38
rs527557219 Chr.12: 19518040-19518040 on Build GRCh38
rs368469203 Chr.12: 19518069-19518069 on Build GRCh38
rs779995362 Chr.12: 19518155-19518155 on Build GRCh38
rs751573497 Chr.12: 19518075-19518075 on Build GRCh38

View Full Product Details