Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48918330-48918577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC65
Entrez Gene ID
85478
Gene Name
coiled-coil domain containing 65
Gene Aliases
CFAP250, CILD27, DRC2, FAP250, NYD-SP28
UniGene
Hs.512805
Gene Chromosome Location
Chr.12: 48904110-48921576 on Build GRCh38
Transcripts
NM_001286957.1 NM_033124.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4575434 c.747G>A p.V249V Chr.12: 48918412-48918412 on Build GRCh38
4615927 c.786_787insA p.I265fs*21 Chr.12: 48918451-48918452 on Build GRCh38
7092657 c.771G>T p.E257D Chr.12: 48918436-48918436 on Build GRCh38
4615928 c.786_787insA p.I265fs*21 Chr.12: 48918451-48918452 on Build GRCh38
4425302 c.716A>G p.D239G Chr.12: 48918381-48918381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748165696 Chr.12: 48918486-48918486 on Build GRCh38
rs750142586 Chr.12: 48918362-48918362 on Build GRCh38
rs529989062 Chr.12: 48918514-48918514 on Build GRCh38
rs755040452 Chr.12: 48918524-48918525 on Build GRCh38
rs778911045 Chr.12: 48918377-48918377 on Build GRCh38
rs775941317 Chr.12: 48918416-48918416 on Build GRCh38
rs201132346 Chr.12: 48918412-48918412 on Build GRCh38
rs764695480 Chr.12: 48918436-48918436 on Build GRCh38
rs755717773 Chr.12: 48918458-48918458 on Build GRCh38
rs940607880 Chr.12: 48918553-48918553 on Build GRCh38
rs780152149 Chr.12: 48918373-48918374 on Build GRCh38
rs1003676784 Chr.12: 48918383-48918383 on Build GRCh38
rs771100560 Chr.12: 48918392-48918392 on Build GRCh38
rs751586867 Chr.12: 48918451-48918452 on Build GRCh38
rs759137345 Chr.12: 48918384-48918384 on Build GRCh38
rs143357639 Chr.12: 48918368-48918368 on Build GRCh38
rs754830260 Chr.12: 48918473-48918473 on Build GRCh38
rs199888057 Chr.12: 48918438-48918438 on Build GRCh38
rs749101471 Chr.12: 48918371-48918371 on Build GRCh38
rs749936821 Chr.12: 48918443-48918443 on Build GRCh38
rs372541279 Chr.12: 48918375-48918375 on Build GRCh38
rs148347233 Chr.12: 48918427-48918427 on Build GRCh38
rs368331248 Chr.12: 48918367-48918367 on Build GRCh38
rs976849380 Chr.12: 48918496-48918496 on Build GRCh38
rs757252272 Chr.12: 48918492-48918492 on Build GRCh38
rs776694186 Chr.12: 48918397-48918397 on Build GRCh38
rs759876162 Chr.12: 48918409-48918409 on Build GRCh38
rs531737974 Chr.12: 48918426-48918426 on Build GRCh38
rs767098849 Chr.12: 48918439-48918439 on Build GRCh38
rs547810139 Chr.12: 48918533-48918542 on Build GRCh38
rs150949692 Chr.12: 48918468-48918468 on Build GRCh38
rs745961187 Chr.12: 48918516-48918516 on Build GRCh38
rs748346015 Chr.12: 48918470-48918470 on Build GRCh38

View Full Product Details