Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71851335-71851608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN1L
Entrez Gene ID
342371
Gene Name
ataxin 1 like
Gene Aliases
BOAT, BOAT1
UniGene
Hs.743239
Gene Chromosome Location
Chr.16: 71845991-71857333 on Build GRCh38
Transcripts
NM_001137675.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
973390 c.1849G>T p.E617* Chr.16: 71851589-71851589 on Build GRCh38
973389 c.1624G>A p.E542K Chr.16: 71851364-71851364 on Build GRCh38
7221326 c.1780A>G p.S594G Chr.16: 71851520-71851520 on Build GRCh38
5617974 c.1790C>A p.P597H Chr.16: 71851530-71851530 on Build GRCh38
6024433 c.1807_1808insC p.R605fs*5 Chr.16: 71851547-71851548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050105146 Chr.16: 71851415-71851415 on Build GRCh38
rs750587081 Chr.16: 71851495-71851495 on Build GRCh38
rs1034718181 Chr.16: 71851552-71851552 on Build GRCh38
rs368301080 Chr.16: 71851416-71851416 on Build GRCh38
rs368130565 Chr.16: 71851529-71851529 on Build GRCh38
rs911754661 Chr.16: 71851456-71851456 on Build GRCh38
rs1004805775 Chr.16: 71851578-71851578 on Build GRCh38
rs761115947 Chr.16: 71851546-71851547 on Build GRCh38
rs774592876 Chr.16: 71851522-71851522 on Build GRCh38
rs1002889027 Chr.16: 71851559-71851559 on Build GRCh38
rs933056015 Chr.16: 71851359-71851359 on Build GRCh38
rs1020278921 Chr.16: 71851535-71851535 on Build GRCh38
rs893734086 Chr.16: 71851526-71851526 on Build GRCh38
rs758910069 Chr.16: 71851545-71851545 on Build GRCh38
rs935147566 Chr.16: 71851472-71851472 on Build GRCh38
rs778138043 Chr.16: 71851551-71851551 on Build GRCh38
rs951133490 Chr.16: 71851567-71851567 on Build GRCh38
rs754024732 Chr.16: 71851524-71851524 on Build GRCh38
rs61747556 Chr.16: 71851530-71851530 on Build GRCh38
rs548910166 Chr.16: 71851579-71851579 on Build GRCh38
rs775793808 Chr.16: 71851357-71851358 on Build GRCh38
rs746577943 Chr.16: 71851554-71851554 on Build GRCh38
rs904865804 Chr.16: 71851531-71851531 on Build GRCh38
rs936479712 Chr.16: 71851500-71851500 on Build GRCh38
rs770317159 Chr.16: 71851572-71851572 on Build GRCh38
rs934099733 Chr.16: 71851420-71851420 on Build GRCh38
rs190299416 Chr.16: 71851540-71851540 on Build GRCh38
rs767447331 Chr.16: 71851498-71851498 on Build GRCh38
rs550606040 Chr.16: 71851451-71851451 on Build GRCh38
rs372403992 Chr.16: 71851357-71851357 on Build GRCh38
rs1051180135 Chr.16: 71851432-71851432 on Build GRCh38
rs766669613 Chr.16: 71851521-71851521 on Build GRCh38
rs61747555 Chr.16: 71851520-71851520 on Build GRCh38
rs199683333 Chr.16: 71851427-71851427 on Build GRCh38

View Full Product Details