Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 16840984-16841243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUBN
Entrez Gene ID
8029
Gene Name
cubilin
Gene Aliases
IFCR, MGA1, gp280
UniGene
Hs.166206
Gene Chromosome Location
Chr.10: 16823966-17130492 on Build GRCh38
Transcripts
NM_001081.3 XM_011519709.2 XM_011519710.2 XM_011519711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3978326 c.9696G>T p.L3232F Chr.10: 16841015-16841015 on Build GRCh38
1347097 c.9667T>C p.Y3223H Chr.10: 16841044-16841044 on Build GRCh38
683785 c.9717C>G p.S3239S Chr.10: 16840994-16840994 on Build GRCh38
5014894 c.9725C>A p.P3242H Chr.10: 16840986-16840986 on Build GRCh38
916769 c.9690G>A p.A3230A Chr.10: 16841021-16841021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018187017 Chr.10: 16841120-16841120 on Build GRCh38
rs143533714 Chr.10: 16841044-16841044 on Build GRCh38
rs372958337 Chr.10: 16841037-16841037 on Build GRCh38
rs773439837 Chr.10: 16841030-16841030 on Build GRCh38
rs976583526 Chr.10: 16841141-16841141 on Build GRCh38
rs370784621 Chr.10: 16841006-16841006 on Build GRCh38
rs763817461 Chr.10: 16841075-16841075 on Build GRCh38
rs756742695 Chr.10: 16841007-16841007 on Build GRCh38
rs762325028 Chr.10: 16841021-16841021 on Build GRCh38
rs769318609 Chr.10: 16841055-16841055 on Build GRCh38
rs756898936 Chr.10: 16841091-16841091 on Build GRCh38
rs868810367 Chr.10: 16841107-16841107 on Build GRCh38
rs770456995 Chr.10: 16841063-16841066 on Build GRCh38
rs372599172 Chr.10: 16841045-16841045 on Build GRCh38
rs151308942 Chr.10: 16841118-16841118 on Build GRCh38
rs1025869715 Chr.10: 16841142-16841142 on Build GRCh38
rs187147305 Chr.10: 16841117-16841117 on Build GRCh38
rs780748417 Chr.10: 16841095-16841095 on Build GRCh38
rs893429695 Chr.10: 16841102-16841102 on Build GRCh38
rs565583251 Chr.10: 16841139-16841139 on Build GRCh38
rs752637481 Chr.10: 16841065-16841065 on Build GRCh38
rs139511152 Chr.10: 16841022-16841022 on Build GRCh38
rs150890299 Chr.10: 16841010-16841010 on Build GRCh38
rs765104843 Chr.10: 16841057-16841057 on Build GRCh38
rs951512784 Chr.10: 16841155-16841155 on Build GRCh38
rs145383974 Chr.10: 16841056-16841056 on Build GRCh38
rs758174278 Chr.10: 16841070-16841070 on Build GRCh38
rs780615051 Chr.10: 16841008-16841008 on Build GRCh38
rs773892841 Chr.10: 16841150-16841150 on Build GRCh38
rs751202387 Chr.10: 16841077-16841077 on Build GRCh38
rs984571664 Chr.10: 16841198-16841198 on Build GRCh38
rs1032009893 Chr.10: 16841083-16841083 on Build GRCh38

View Full Product Details