Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61879854-61880127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS3
Entrez Gene ID
3995
Gene Name
fatty acid desaturase 3
Gene Aliases
CYB5RP, LLCDL3
UniGene
Hs.21765
Gene Chromosome Location
Chr.11: 61873523-61892224 on Build GRCh38
Transcripts
NM_021727.4 XM_011545023.2 XM_017017723.1 XM_017017724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1355280 c.254G>A p.R85H Chr.11: 61880111-61880111 on Build GRCh38
6657929 c.238G>T p.D80Y Chr.11: 61880127-61880127 on Build GRCh38
5970237 c.293C>T p.P98L Chr.11: 61880072-61880072 on Build GRCh38
6657930 c.238G>T p.D80Y Chr.11: 61880127-61880127 on Build GRCh38
4903275 c.307C>T p.Q103* Chr.11: 61880058-61880058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747080847 Chr.11: 61880099-61880099 on Build GRCh38
rs957812527 Chr.11: 61879880-61879880 on Build GRCh38
rs934896882 Chr.11: 61879884-61879884 on Build GRCh38
rs547830697 Chr.11: 61879953-61879953 on Build GRCh38
rs781377821 Chr.11: 61880103-61880103 on Build GRCh38
rs188696017 Chr.11: 61879938-61879938 on Build GRCh38
rs570760086 Chr.11: 61880034-61880034 on Build GRCh38
rs966519520 Chr.11: 61879919-61879919 on Build GRCh38
rs918722904 Chr.11: 61880057-61880057 on Build GRCh38
rs766894921 Chr.11: 61879998-61879998 on Build GRCh38
rs1053815361 Chr.11: 61879891-61879891 on Build GRCh38
rs990861591 Chr.11: 61879967-61879967 on Build GRCh38
rs559059824 Chr.11: 61879931-61879931 on Build GRCh38
rs866390788 Chr.11: 61880095-61880095 on Build GRCh38
rs753614899 Chr.11: 61880021-61880021 on Build GRCh38
rs927937825 Chr.11: 61879946-61879946 on Build GRCh38
rs752511844 Chr.11: 61880049-61880049 on Build GRCh38
rs777666692 Chr.11: 61880098-61880098 on Build GRCh38
rs757347000 Chr.11: 61880100-61880100 on Build GRCh38
rs191501106 Chr.11: 61880022-61880022 on Build GRCh38
rs1051778381 Chr.11: 61880072-61880072 on Build GRCh38
rs368351930 Chr.11: 61880071-61880071 on Build GRCh38
rs746014083 Chr.11: 61880011-61880011 on Build GRCh38
rs374211652 Chr.11: 61880030-61880030 on Build GRCh38
rs772796842 Chr.11: 61879999-61879999 on Build GRCh38
rs760148251 Chr.11: 61880010-61880010 on Build GRCh38
rs115905177 Chr.11: 61879910-61879910 on Build GRCh38
rs174635 Chr.11: 61879955-61879955 on Build GRCh38
rs909958602 Chr.11: 61879974-61879974 on Build GRCh38
rs174634 Chr.11: 61879915-61879915 on Build GRCh38

View Full Product Details