Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96841999-96842244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD23
Entrez Gene ID
200539
Gene Name
ankyrin repeat domain 23
Gene Aliases
DARP, MARP3
UniGene
Hs.643430
Gene Chromosome Location
Chr.2: 96837914-96844021 on Build GRCh38
Transcripts
NM_144994.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5645695 c.205C>G p.L69V Chr.2: 96842155-96842155 on Build GRCh38
1023999 c.175-1G>A p.? Chr.2: 96842186-96842186 on Build GRCh38
4759137 c.268G>C p.V90L Chr.2: 96842092-96842092 on Build GRCh38
6269178 c.256C>A p.L86M Chr.2: 96842104-96842104 on Build GRCh38
1410102 c.276_277insC p.R93fs*4 Chr.2: 96842083-96842084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773404418 Chr.2: 96842033-96842033 on Build GRCh38
rs755788231 Chr.2: 96842077-96842077 on Build GRCh38
rs552835249 Chr.2: 96842126-96842126 on Build GRCh38
rs747154444 Chr.2: 96842045-96842045 on Build GRCh38
rs776581688 Chr.2: 96842096-96842096 on Build GRCh38
rs767047953 Chr.2: 96842150-96842150 on Build GRCh38
rs150321775 Chr.2: 96842170-96842170 on Build GRCh38
rs778384595 Chr.2: 96842086-96842086 on Build GRCh38
rs759694604 Chr.2: 96842057-96842057 on Build GRCh38
rs1044751136 Chr.2: 96842081-96842081 on Build GRCh38
rs771106274 Chr.2: 96842083-96842084 on Build GRCh38
rs762693200 Chr.2: 96842116-96842116 on Build GRCh38
rs148240041 Chr.2: 96842121-96842121 on Build GRCh38
rs202222596 Chr.2: 96842106-96842106 on Build GRCh38
rs773775937 Chr.2: 96842023-96842024 on Build GRCh38
rs920969428 Chr.2: 96842143-96842143 on Build GRCh38
rs775332867 Chr.2: 96842063-96842063 on Build GRCh38
rs761744967 Chr.2: 96842178-96842178 on Build GRCh38
rs774456442 Chr.2: 96842084-96842084 on Build GRCh38
rs141023516 Chr.2: 96842107-96842107 on Build GRCh38
rs761660231 Chr.2: 96842137-96842137 on Build GRCh38
rs749957437 Chr.2: 96842151-96842151 on Build GRCh38
rs763223465 Chr.2: 96842025-96842026 on Build GRCh38
rs779406139 Chr.2: 96842084-96842084 on Build GRCh38
rs779423681 Chr.2: 96842023-96842023 on Build GRCh38
rs747487070 Chr.2: 96842209-96842209 on Build GRCh38
rs778300385 Chr.2: 96842207-96842207 on Build GRCh38
rs759492213 Chr.2: 96842084-96842085 on Build GRCh38
rs750023979 Chr.2: 96842074-96842074 on Build GRCh38
rs764056715 Chr.2: 96842065-96842065 on Build GRCh38
rs551667739 Chr.2: 96842061-96842061 on Build GRCh38
rs939741820 Chr.2: 96842205-96842205 on Build GRCh38
rs528045312 Chr.2: 96842049-96842049 on Build GRCh38
rs896232701 Chr.2: 96842076-96842076 on Build GRCh38
rs772765570 Chr.2: 96842030-96842030 on Build GRCh38
rs374100204 Chr.2: 96842066-96842066 on Build GRCh38
rs370445494 Chr.2: 96842032-96842032 on Build GRCh38
rs747492584 Chr.2: 96842092-96842092 on Build GRCh38
rs1038504463 Chr.2: 96842221-96842221 on Build GRCh38
rs767608781 Chr.2: 96842090-96842091 on Build GRCh38
rs766950020 Chr.2: 96842068-96842068 on Build GRCh38
rs758944303 Chr.2: 96842085-96842085 on Build GRCh38
rs993742185 Chr.2: 96842193-96842193 on Build GRCh38
rs559278501 Chr.2: 96842046-96842046 on Build GRCh38
rs377092322 Chr.2: 96842024-96842024 on Build GRCh38
rs898769107 Chr.2: 96842036-96842036 on Build GRCh38
rs113774254 Chr.2: 96842099-96842099 on Build GRCh38
rs758787839 Chr.2: 96842184-96842184 on Build GRCh38
rs753718887 Chr.2: 96842143-96842144 on Build GRCh38
rs201277049 Chr.2: 96842175-96842175 on Build GRCh38
rs757773427 Chr.2: 96842213-96842213 on Build GRCh38
rs201334613 Chr.2: 96842169-96842169 on Build GRCh38
rs1006130993 Chr.2: 96842165-96842165 on Build GRCh38
rs144150866 Chr.2: 96842090-96842090 on Build GRCh38

View Full Product Details