Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26596838-26597091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABT1
Entrez Gene ID
29777
Gene Name
activator of basal transcription 1
Gene Aliases
hABT1
UniGene
Hs.254406
Gene Chromosome Location
Chr.6: 26596943-26600050 on Build GRCh38
Transcripts
NM_013375.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1621305 c.2T>C p.M1T Chr.6: 26596984-26596984 on Build GRCh38
3861018 c.109T>C p.C37R Chr.6: 26597091-26597091 on Build GRCh38
3715373 c.17C>A p.S6* Chr.6: 26596999-26596999 on Build GRCh38
6173382 c.10G>T p.E4* Chr.6: 26596992-26596992 on Build GRCh38
1442692 c.100G>T p.E34* Chr.6: 26597082-26597082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782014861 Chr.6: 26596983-26596983 on Build GRCh38
rs530320253 Chr.6: 26596858-26596858 on Build GRCh38
rs782373630 Chr.6: 26597014-26597014 on Build GRCh38
rs782768842 Chr.6: 26596991-26596991 on Build GRCh38
rs781816337 Chr.6: 26597063-26597063 on Build GRCh38
rs782753754 Chr.6: 26597069-26597069 on Build GRCh38
rs782277922 Chr.6: 26596962-26596962 on Build GRCh38
rs782050243 Chr.6: 26596984-26596984 on Build GRCh38
rs569271744 Chr.6: 26596964-26596964 on Build GRCh38
rs148186414 Chr.6: 26597049-26597049 on Build GRCh38
rs573149951 Chr.6: 26596949-26596949 on Build GRCh38
rs782191822 Chr.6: 26596968-26596968 on Build GRCh38
rs539980770 Chr.6: 26596933-26596933 on Build GRCh38
rs782730484 Chr.6: 26596985-26596985 on Build GRCh38
rs978132300 Chr.6: 26597036-26597036 on Build GRCh38
rs539180441 Chr.6: 26596946-26596946 on Build GRCh38
rs563288722 Chr.6: 26597001-26597001 on Build GRCh38
rs536915270 Chr.6: 26597035-26597035 on Build GRCh38
rs115596326 Chr.6: 26596869-26596869 on Build GRCh38
rs781955349 Chr.6: 26596942-26596943 on Build GRCh38
rs782151876 Chr.6: 26596972-26596972 on Build GRCh38
rs1022530159 Chr.6: 26597026-26597026 on Build GRCh38
rs782230542 Chr.6: 26597011-26597011 on Build GRCh38
rs371015299 Chr.6: 26597056-26597056 on Build GRCh38
rs149473775 Chr.6: 26597018-26597018 on Build GRCh38
rs782140804 Chr.6: 26597043-26597043 on Build GRCh38
rs781995841 Chr.6: 26597042-26597042 on Build GRCh38
rs138003871 Chr.6: 26596982-26596982 on Build GRCh38
rs782603117 Chr.6: 26597010-26597010 on Build GRCh38
rs782108390 Chr.6: 26596983-26596984 on Build GRCh38
rs781937269 Chr.6: 26597029-26597029 on Build GRCh38
rs782543304 Chr.6: 26596998-26596998 on Build GRCh38
rs782442053 Chr.6: 26597003-26597003 on Build GRCh38
rs782125618 Chr.6: 26596936-26596936 on Build GRCh38
rs755965085 Chr.6: 26596966-26596966 on Build GRCh38
rs782354423 Chr.6: 26597039-26597039 on Build GRCh38
rs367775660 Chr.6: 26597046-26597046 on Build GRCh38
rs782771835 Chr.6: 26597063-26597063 on Build GRCh38
rs781847204 Chr.6: 26596958-26596958 on Build GRCh38
rs968684075 Chr.6: 26597034-26597034 on Build GRCh38
rs377021170 Chr.6: 26596978-26596978 on Build GRCh38
rs782679594 Chr.6: 26596999-26596999 on Build GRCh38
rs782489929 Chr.6: 26597067-26597067 on Build GRCh38
rs1005200017 Chr.6: 26596975-26596975 on Build GRCh38
rs781822353 Chr.6: 26596988-26596988 on Build GRCh38
rs373069771 Chr.6: 26596934-26596934 on Build GRCh38
rs573455866 Chr.6: 26597072-26597072 on Build GRCh38
rs976133749 Chr.6: 26597064-26597064 on Build GRCh38
rs782100852 Chr.6: 26596990-26596990 on Build GRCh38
rs991099116 Chr.6: 26597025-26597025 on Build GRCh38
rs368920667 Chr.6: 26596945-26596945 on Build GRCh38
rs782358880 Chr.6: 26596896-26596896 on Build GRCh38
rs782335352 Chr.6: 26596970-26596970 on Build GRCh38

View Full Product Details