Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28503017-28503252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD3
Entrez Gene ID
7976
Gene Name
frizzled class receptor 3
Gene Aliases
Fz-3
UniGene
Hs.40735
Gene Chromosome Location
Chr.8: 28494205-28574268 on Build GRCh38
Transcripts
NM_017412.3 NM_145866.1 XM_017013842.1 XM_017013843.1 XM_017013844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1098899 c.33G>A p.W11* Chr.8: 28503046-28503046 on Build GRCh38
7423509 c.99G>C p.L33F Chr.8: 28503112-28503112 on Build GRCh38
1098900 c.40A>G p.T14A Chr.8: 28503053-28503053 on Build GRCh38
351770 c.64G>T p.G22W Chr.8: 28503077-28503077 on Build GRCh38
1635748 c.95C>A p.T32N Chr.8: 28503108-28503108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192635124 Chr.8: 28503184-28503184 on Build GRCh38
rs146921825 Chr.8: 28503062-28503062 on Build GRCh38
rs532933401 Chr.8: 28503216-28503216 on Build GRCh38
rs771902243 Chr.8: 28503161-28503161 on Build GRCh38
rs983860869 Chr.8: 28503088-28503088 on Build GRCh38
rs751811704 Chr.8: 28503221-28503221 on Build GRCh38
rs745487080 Chr.8: 28503136-28503136 on Build GRCh38
rs774073841 Chr.8: 28503133-28503133 on Build GRCh38
rs113797404 Chr.8: 28503172-28503172 on Build GRCh38
rs943517542 Chr.8: 28503165-28503165 on Build GRCh38
rs368293589 Chr.8: 28503217-28503217 on Build GRCh38
rs766547919 Chr.8: 28503219-28503219 on Build GRCh38
rs775307553 Chr.8: 28503163-28503163 on Build GRCh38
rs771488481 Chr.8: 28503102-28503102 on Build GRCh38
rs762446179 Chr.8: 28503209-28503209 on Build GRCh38
rs777505254 Chr.8: 28503061-28503061 on Build GRCh38
rs1055065673 Chr.8: 28503054-28503054 on Build GRCh38
rs61754565 Chr.8: 28503197-28503197 on Build GRCh38
rs777708535 Chr.8: 28503104-28503104 on Build GRCh38
rs770335409 Chr.8: 28503115-28503115 on Build GRCh38
rs375620381 Chr.8: 28503052-28503052 on Build GRCh38
rs749262039 Chr.8: 28503098-28503098 on Build GRCh38
rs762989332 Chr.8: 28503049-28503049 on Build GRCh38
rs764528480 Chr.8: 28503185-28503185 on Build GRCh38

View Full Product Details