Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57202432-57202675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP7
Entrez Gene ID
655
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP-1
UniGene
Hs.473163
Gene Chromosome Location
Chr.20: 57168753-57266651 on Build GRCh38
Transcripts
NM_001719.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6274848 c.653C>A p.A218D Chr.20: 57202582-57202582 on Build GRCh38
4607792 c.705G>T p.W235C Chr.20: 57202530-57202530 on Build GRCh38
2154712 c.754C>T p.L252L Chr.20: 57202481-57202481 on Build GRCh38
1028336 c.744G>A p.S248S Chr.20: 57202491-57202491 on Build GRCh38
2764200 c.757G>A p.D253N Chr.20: 57202478-57202478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767362735 Chr.20: 57202656-57202656 on Build GRCh38
rs767615840 Chr.20: 57202475-57202475 on Build GRCh38
rs775359806 Chr.20: 57202551-57202551 on Build GRCh38
rs202084512 Chr.20: 57202516-57202516 on Build GRCh38
rs749814408 Chr.20: 57202535-57202535 on Build GRCh38
rs943506437 Chr.20: 57202638-57202638 on Build GRCh38
rs145527493 Chr.20: 57202492-57202492 on Build GRCh38
rs112344257 Chr.20: 57202483-57202483 on Build GRCh38
rs368039601 Chr.20: 57202518-57202518 on Build GRCh38
rs758218504 Chr.20: 57202504-57202504 on Build GRCh38
rs773246134 Chr.20: 57202581-57202581 on Build GRCh38
rs116720786 Chr.20: 57202517-57202517 on Build GRCh38
rs760614323 Chr.20: 57202578-57202578 on Build GRCh38
rs114671049 Chr.20: 57202628-57202628 on Build GRCh38
rs534952030 Chr.20: 57202653-57202653 on Build GRCh38
rs746092842 Chr.20: 57202550-57202550 on Build GRCh38
rs775789240 Chr.20: 57202561-57202561 on Build GRCh38
rs759706614 Chr.20: 57202469-57202469 on Build GRCh38
rs74573730 Chr.20: 57202552-57202552 on Build GRCh38
rs752403942 Chr.20: 57202601-57202601 on Build GRCh38
rs372592843 Chr.20: 57202464-57202464 on Build GRCh38
rs535901102 Chr.20: 57202582-57202582 on Build GRCh38
rs368736590 Chr.20: 57202452-57202452 on Build GRCh38
rs771242712 Chr.20: 57202544-57202544 on Build GRCh38
rs772253049 Chr.20: 57202560-57202560 on Build GRCh38
rs141957513 Chr.20: 57202491-57202491 on Build GRCh38
rs779335170 Chr.20: 57202617-57202617 on Build GRCh38
rs764881969 Chr.20: 57202542-57202542 on Build GRCh38
rs528012771 Chr.20: 57202463-57202463 on Build GRCh38
rs757821410 Chr.20: 57202606-57202606 on Build GRCh38
rs367817895 Chr.20: 57202646-57202646 on Build GRCh38
rs763025993 Chr.20: 57202587-57202587 on Build GRCh38
rs138140607 Chr.20: 57202618-57202618 on Build GRCh38
rs370126479 Chr.20: 57202594-57202594 on Build GRCh38
rs774630143 Chr.20: 57202546-57202546 on Build GRCh38
rs751368146 Chr.20: 57202591-57202591 on Build GRCh38
rs570170347 Chr.20: 57202522-57202522 on Build GRCh38
rs1000981409 Chr.20: 57202615-57202615 on Build GRCh38
rs767011450 Chr.20: 57202595-57202595 on Build GRCh38
rs905272153 Chr.20: 57202614-57202614 on Build GRCh38
rs142693632 Chr.20: 57202623-57202623 on Build GRCh38
rs775526020 Chr.20: 57202627-57202627 on Build GRCh38
rs752779943 Chr.20: 57202480-57202480 on Build GRCh38
rs780738237 Chr.20: 57202519-57202519 on Build GRCh38
rs777292866 Chr.20: 57202602-57202602 on Build GRCh38
rs755625837 Chr.20: 57202526-57202526 on Build GRCh38
rs768827500 Chr.20: 57202630-57202630 on Build GRCh38
rs200781040 Chr.20: 57202655-57202655 on Build GRCh38
rs778190724 Chr.20: 57202527-57202527 on Build GRCh38
rs764292927 Chr.20: 57202579-57202579 on Build GRCh38
rs201253232 Chr.20: 57202647-57202647 on Build GRCh38

View Full Product Details