Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196902292-196902553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR4
Entrez Gene ID
10877
Gene Name
complement factor H related 4
Gene Aliases
CFHL4, FHR-4, FHR4
UniGene
Hs.710100
Gene Chromosome Location
Chr.1: 196888014-196918911 on Build GRCh38
Transcripts
NM_001201550.2 NM_001201551.1 NM_006684.4 XM_006711129.3 XM_017000110.1 XM_017000111.1 XM_017000112.1 XM_017000113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4958410 c.123A>G p.L41L Chr.1: 196902485-196902485 on Build GRCh38
7094327 c.191G>A p.G64E Chr.1: 196902553-196902553 on Build GRCh38
6060109 c.71G>T p.C24F Chr.1: 196902430-196902430 on Build GRCh38
1727394 c.87A>T p.Q29H Chr.1: 196902449-196902449 on Build GRCh38
5590184 c.59-1G>A p.? Chr.1: 196902417-196902417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559968296 Chr.1: 196902370-196902370 on Build GRCh38
rs754348652 Chr.1: 196902443-196902443 on Build GRCh38
rs370481486 Chr.1: 196902450-196902450 on Build GRCh38
rs761343811 Chr.1: 196902410-196902410 on Build GRCh38
rs774236421 Chr.1: 196902403-196902403 on Build GRCh38
rs769627887 Chr.1: 196902383-196902383 on Build GRCh38
rs368555895 Chr.1: 196902501-196902501 on Build GRCh38
rs749052570 Chr.1: 196902512-196902512 on Build GRCh38
rs199547603 Chr.1: 196902480-196902480 on Build GRCh38
rs774033155 Chr.1: 196902407-196902407 on Build GRCh38
rs374652211 Chr.1: 196902520-196902520 on Build GRCh38
rs761008403 Chr.1: 196902433-196902433 on Build GRCh38
rs370363594 Chr.1: 196902396-196902396 on Build GRCh38
rs560734727 Chr.1: 196902426-196902426 on Build GRCh38
rs779923402 Chr.1: 196902506-196902506 on Build GRCh38
rs1014176613 Chr.1: 196902334-196902334 on Build GRCh38
rs957453338 Chr.1: 196902444-196902444 on Build GRCh38
rs200339924 Chr.1: 196902477-196902477 on Build GRCh38
rs78951215 Chr.1: 196902518-196902518 on Build GRCh38
rs369421996 Chr.1: 196902327-196902327 on Build GRCh38
rs760090260 Chr.1: 196902404-196902406 on Build GRCh38
rs897557090 Chr.1: 196902365-196902365 on Build GRCh38
rs761471251 Chr.1: 196902522-196902522 on Build GRCh38
rs762927352 Chr.1: 196902386-196902386 on Build GRCh38
rs766731028 Chr.1: 196902436-196902436 on Build GRCh38
rs779352004 Chr.1: 196902452-196902452 on Build GRCh38
rs200977143 Chr.1: 196902478-196902478 on Build GRCh38
rs771029005 Chr.1: 196902368-196902369 on Build GRCh38
rs1024641945 Chr.1: 196902367-196902367 on Build GRCh38
rs775329784 Chr.1: 196902384-196902384 on Build GRCh38
rs116125364 Chr.1: 196902404-196902404 on Build GRCh38
rs929065108 Chr.1: 196902378-196902378 on Build GRCh38
rs772311435 Chr.1: 196902368-196902368 on Build GRCh38
rs745663371 Chr.1: 196902371-196902371 on Build GRCh38
rs78624607 Chr.1: 196902421-196902421 on Build GRCh38
rs202234955 Chr.1: 196902487-196902487 on Build GRCh38
rs138792300 Chr.1: 196902462-196902462 on Build GRCh38
rs767296457 Chr.1: 196902413-196902413 on Build GRCh38

View Full Product Details