Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167553319-167553562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREG1
Entrez Gene ID
8804
Gene Name
cellular repressor of E1A stimulated genes 1
Gene Aliases
CREG
UniGene
Hs.5710
Gene Chromosome Location
Chr.1: 167541013-167553819 on Build GRCh38
Transcripts
NM_003851.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6560962 c.315C>G p.F105L Chr.1: 167553427-167553427 on Build GRCh38
5673372 c.215C>T p.A72V Chr.1: 167553527-167553527 on Build GRCh38
1172060 c.335T>G p.L112R Chr.1: 167553407-167553407 on Build GRCh38
6541203 c.190C>A p.H64N Chr.1: 167553552-167553552 on Build GRCh38
1172061 c.333G>C p.Q111H Chr.1: 167553409-167553409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892449502 Chr.1: 167553378-167553378 on Build GRCh38
rs1004929912 Chr.1: 167553406-167553410 on Build GRCh38
rs750704366 Chr.1: 167553417-167553417 on Build GRCh38
rs963561465 Chr.1: 167553425-167553425 on Build GRCh38
rs962004804 Chr.1: 167553481-167553481 on Build GRCh38
rs923990934 Chr.1: 167553476-167553476 on Build GRCh38
rs969186449 Chr.1: 167553536-167553536 on Build GRCh38
rs781036042 Chr.1: 167553487-167553487 on Build GRCh38
rs780115939 Chr.1: 167553448-167553448 on Build GRCh38
rs543544332 Chr.1: 167553517-167553517 on Build GRCh38
rs1015892042 Chr.1: 167553479-167553479 on Build GRCh38
rs1052333157 Chr.1: 167553355-167553355 on Build GRCh38
rs754251882 Chr.1: 167553357-167553357 on Build GRCh38
rs565016025 Chr.1: 167553519-167553519 on Build GRCh38
rs900337337 Chr.1: 167553361-167553361 on Build GRCh38
rs754935394 Chr.1: 167553466-167553466 on Build GRCh38
rs899922736 Chr.1: 167553416-167553416 on Build GRCh38
rs989854698 Chr.1: 167553489-167553489 on Build GRCh38
rs915238501 Chr.1: 167553501-167553501 on Build GRCh38
rs997301436 Chr.1: 167553365-167553365 on Build GRCh38
rs997399081 Chr.1: 167553433-167553433 on Build GRCh38
rs1051939621 Chr.1: 167553367-167553367 on Build GRCh38
rs1023797901 Chr.1: 167553451-167553451 on Build GRCh38
rs776820168 Chr.1: 167553514-167553514 on Build GRCh38
rs1012442489 Chr.1: 167553450-167553450 on Build GRCh38
rs765631682 Chr.1: 167553398-167553398 on Build GRCh38
rs1038953438 Chr.1: 167553515-167553515 on Build GRCh38
rs34590235 Chr.1: 167553483-167553484 on Build GRCh38
rs757603428 Chr.1: 167553382-167553382 on Build GRCh38
rs1041518946 Chr.1: 167553406-167553406 on Build GRCh38
rs970851079 Chr.1: 167553467-167553467 on Build GRCh38
rs982246929 Chr.1: 167553473-167553473 on Build GRCh38
rs747054186 Chr.1: 167553460-167553460 on Build GRCh38
rs369733737 Chr.1: 167553442-167553442 on Build GRCh38

View Full Product Details