Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 12138911-12139142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMPD4
Entrez Gene ID
9758
Gene Name
FERM and PDZ domain containing 4
Gene Aliases
PDZD10, PDZK10
UniGene
Hs.657507
Gene Chromosome Location
Chr.X: 12137329-12724523 on Build GRCh38
Transcripts
NM_014728.3 XM_011545613.2 XM_017029983.1 XM_017029986.1

SNP Detail

SNP ID Chromosome Location
rs748919873 Chr.X: 12138949-12138949 on Build GRCh38
rs374838344 Chr.X: 12138956-12138956 on Build GRCh38
rs770446012 Chr.X: 12139066-12139066 on Build GRCh38
rs370429800 Chr.X: 12138937-12138937 on Build GRCh38
rs779208984 Chr.X: 12138934-12138934 on Build GRCh38
rs745864656 Chr.X: 12138958-12138958 on Build GRCh38
rs770861698 Chr.X: 12138952-12138952 on Build GRCh38
rs140506521 Chr.X: 12138941-12138941 on Build GRCh38
rs777325845 Chr.X: 12138946-12138946 on Build GRCh38
rs776404518 Chr.X: 12139119-12139119 on Build GRCh38
rs988994228 Chr.X: 12139083-12139083 on Build GRCh38
rs926151782 Chr.X: 12139031-12139031 on Build GRCh38
rs773320993 Chr.X: 12139063-12139063 on Build GRCh38
rs765318761 Chr.X: 12139008-12139008 on Build GRCh38
rs746710388 Chr.X: 12139057-12139057 on Build GRCh38
rs772812169 Chr.X: 12139025-12139025 on Build GRCh38
rs377747533 Chr.X: 12138997-12138997 on Build GRCh38
rs776821548 Chr.X: 12138986-12138986 on Build GRCh38
rs148929583 Chr.X: 12138998-12138998 on Build GRCh38
rs916905587 Chr.X: 12139090-12139090 on Build GRCh38
rs772121141 Chr.X: 12138980-12138980 on Build GRCh38
rs1008281011 Chr.X: 12139047-12139047 on Build GRCh38

View Full Product Details