Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240870583-240870850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGXT
Entrez Gene ID
189
Gene Name
alanine-glyoxylate aminotransferase
Gene Aliases
AGT, AGT1, AGXT1, PH1, SPAT, SPT, TLH6
UniGene
Hs.144567
Gene Chromosome Location
Chr.2: 240868745-240879119 on Build GRCh38
Transcripts
NM_000030.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354850 c.374C>T p.P125L Chr.2: 240870659-240870659 on Build GRCh38
7354852 c.395G>C p.G132A Chr.2: 240870680-240870680 on Build GRCh38
5396642 c.412G>A p.E138K Chr.2: 240870697-240870697 on Build GRCh38
5725388 c.421G>A p.E141K Chr.2: 240870706-240870706 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs117043148 Chr.2: 240870737-240870737 on Build GRCh38
rs777932746 Chr.2: 240870636-240870636 on Build GRCh38
rs564305705 Chr.2: 240870768-240870768 on Build GRCh38
rs1004510369 Chr.2: 240870719-240870719 on Build GRCh38
rs754716643 Chr.2: 240870660-240870660 on Build GRCh38
rs146723289 Chr.2: 240870717-240870717 on Build GRCh38
rs946054636 Chr.2: 240870783-240870783 on Build GRCh38
rs112910630 Chr.2: 240870710-240870710 on Build GRCh38
rs958519122 Chr.2: 240870745-240870745 on Build GRCh38
rs977482622 Chr.2: 240870787-240870787 on Build GRCh38
rs180177214 Chr.2: 240870694-240870694 on Build GRCh38
rs180177217 Chr.2: 240870708-240870708 on Build GRCh38
rs574091775 Chr.2: 240870720-240870720 on Build GRCh38
rs180177212 Chr.2: 240870670-240870670 on Build GRCh38
rs758789348 Chr.2: 240870621-240870621 on Build GRCh38
rs545708737 Chr.2: 240870765-240870765 on Build GRCh38
rs528216770 Chr.2: 240870802-240870802 on Build GRCh38
rs180177211 Chr.2: 240870656-240870656 on Build GRCh38
rs1012633500 Chr.2: 240870624-240870624 on Build GRCh38
rs540334603 Chr.2: 240870803-240870803 on Build GRCh38
rs368013857 Chr.2: 240870641-240870641 on Build GRCh38
rs866739000 Chr.2: 240870721-240870721 on Build GRCh38
rs962969778 Chr.2: 240870722-240870722 on Build GRCh38
rs766935712 Chr.2: 240870729-240870729 on Build GRCh38
rs894301533 Chr.2: 240870620-240870620 on Build GRCh38
rs551537424 Chr.2: 240870699-240870699 on Build GRCh38
rs117357855 Chr.2: 240870753-240870753 on Build GRCh38
rs575985084 Chr.2: 240870659-240870659 on Build GRCh38
rs781391807 Chr.2: 240870650-240870650 on Build GRCh38
rs180177210 Chr.2: 240870649-240870649 on Build GRCh38
rs376589297 Chr.2: 240870738-240870738 on Build GRCh38
rs912550522 Chr.2: 240870801-240870801 on Build GRCh38
rs914468289 Chr.2: 240870770-240870770 on Build GRCh38
rs180177216 Chr.2: 240870744-240870744 on Build GRCh38
rs944160082 Chr.2: 240870813-240870813 on Build GRCh38
rs971207168 Chr.2: 240870725-240870725 on Build GRCh38
rs573283978 Chr.2: 240870609-240870609 on Build GRCh38
rs180177215 Chr.2: 240870701-240870703 on Build GRCh38

View Full Product Details