Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37819229-37819502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF10
Entrez Gene ID
79269
Gene Name
DDB1 and CUL4 associated factor 10
Gene Aliases
WDR32
UniGene
Hs.118394
Gene Chromosome Location
Chr.9: 37800554-37867669 on Build GRCh38
Transcripts
NM_001286810.1 NM_024345.4 XM_005251577.3 XM_005251578.3 XM_017015128.1 XM_017015129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
487468 c.563A>G p.E188G Chr.9: 37819311-37819311 on Build GRCh38
5186737 c.609delA p.T205fs*10 Chr.9: 37819357-37819357 on Build GRCh38
1109071 c.546G>A p.V182V Chr.9: 37819294-37819294 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs890609720 Chr.9: 37819470-37819470 on Build GRCh38
rs761148736 Chr.9: 37819270-37819270 on Build GRCh38
rs559850513 Chr.9: 37819412-37819412 on Build GRCh38
rs762313816 Chr.9: 37819313-37819313 on Build GRCh38
rs763798331 Chr.9: 37819411-37819411 on Build GRCh38
rs184522240 Chr.9: 37819449-37819449 on Build GRCh38
rs758384818 Chr.9: 37819440-37819440 on Build GRCh38
rs756001166 Chr.9: 37819404-37819404 on Build GRCh38
rs767242250 Chr.9: 37819361-37819361 on Build GRCh38
rs574415201 Chr.9: 37819343-37819343 on Build GRCh38
rs144082775 Chr.9: 37819303-37819303 on Build GRCh38
rs540246681 Chr.9: 37819348-37819348 on Build GRCh38
rs759010427 Chr.9: 37819351-37819351 on Build GRCh38
rs778954582 Chr.9: 37819429-37819429 on Build GRCh38
rs765675704 Chr.9: 37819342-37819342 on Build GRCh38
rs367880912 Chr.9: 37819438-37819438 on Build GRCh38
rs757278264 Chr.9: 37819414-37819414 on Build GRCh38
rs768890655 Chr.9: 37819281-37819281 on Build GRCh38
rs752272671 Chr.9: 37819385-37819385 on Build GRCh38
rs1019901225 Chr.9: 37819441-37819441 on Build GRCh38
rs192158102 Chr.9: 37819433-37819433 on Build GRCh38

View Full Product Details