Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89583828-89584071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_011523001.2 XM_017023138.1 XM_017023139.1 XM_017023140.1 XM_017023141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
705303 c.677C>T p.S226L Chr.16: 89584047-89584047 on Build GRCh38
705304 c.675C>A p.I225I Chr.16: 89584045-89584045 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762377005 Chr.16: 89584011-89584011 on Build GRCh38
rs986627081 Chr.16: 89583978-89583978 on Build GRCh38
rs141398937 Chr.16: 89583974-89583974 on Build GRCh38
rs897542705 Chr.16: 89583846-89583846 on Build GRCh38
rs576526147 Chr.16: 89583921-89583921 on Build GRCh38
rs941862934 Chr.16: 89583952-89583952 on Build GRCh38
rs917945014 Chr.16: 89583913-89583913 on Build GRCh38
rs979585229 Chr.16: 89583897-89583898 on Build GRCh38
rs772502780 Chr.16: 89584034-89584034 on Build GRCh38
rs886653276 Chr.16: 89583915-89583915 on Build GRCh38
rs774246153 Chr.16: 89584043-89584043 on Build GRCh38
rs770545030 Chr.16: 89583864-89583864 on Build GRCh38
rs408800 Chr.16: 89583963-89583963 on Build GRCh38
rs760774452 Chr.16: 89583884-89583884 on Build GRCh38
rs924916324 Chr.16: 89584012-89584012 on Build GRCh38
rs964595049 Chr.16: 89583885-89583885 on Build GRCh38
rs377030837 Chr.16: 89584000-89584000 on Build GRCh38
rs369886930 Chr.16: 89584037-89584037 on Build GRCh38
rs529539909 Chr.16: 89583999-89583999 on Build GRCh38
rs1016652491 Chr.16: 89583922-89583922 on Build GRCh38
rs925366874 Chr.16: 89583899-89583899 on Build GRCh38
rs974111557 Chr.16: 89583916-89583916 on Build GRCh38
rs373955797 Chr.16: 89584033-89584033 on Build GRCh38
rs993132802 Chr.16: 89583847-89583847 on Build GRCh38
rs917664196 Chr.16: 89584025-89584025 on Build GRCh38
rs186909019 Chr.16: 89583901-89583901 on Build GRCh38
rs139532630 Chr.16: 89584041-89584041 on Build GRCh38
rs1037466253 Chr.16: 89583953-89583953 on Build GRCh38
rs1029926421 Chr.16: 89583856-89583856 on Build GRCh38
rs372418637 Chr.16: 89583982-89583982 on Build GRCh38
rs749539380 Chr.16: 89584014-89584014 on Build GRCh38
rs529101968 Chr.16: 89584047-89584047 on Build GRCh38
rs747643353 Chr.16: 89584036-89584036 on Build GRCh38
rs925064733 Chr.16: 89583860-89583860 on Build GRCh38
rs779193206 Chr.16: 89584021-89584021 on Build GRCh38
rs772894474 Chr.16: 89584038-89584038 on Build GRCh38
rs375204346 Chr.16: 89584035-89584035 on Build GRCh38
rs926555905 Chr.16: 89583942-89583942 on Build GRCh38
rs546278189 Chr.16: 89583909-89583909 on Build GRCh38
rs752736185 Chr.16: 89584049-89584049 on Build GRCh38
rs146394111 Chr.16: 89584010-89584010 on Build GRCh38
rs558284865 Chr.16: 89583918-89583918 on Build GRCh38
rs949399309 Chr.16: 89583929-89583929 on Build GRCh38
rs145150175 Chr.16: 89584048-89584048 on Build GRCh38
rs1018515798 Chr.16: 89583881-89583881 on Build GRCh38
rs376389386 Chr.16: 89584015-89584015 on Build GRCh38
rs1030406384 Chr.16: 89583973-89583973 on Build GRCh38
rs781307228 Chr.16: 89584003-89584003 on Build GRCh38
rs534559082 Chr.16: 89583861-89583861 on Build GRCh38
rs900604604 Chr.16: 89583932-89583932 on Build GRCh38
rs367582915 Chr.16: 89583970-89583970 on Build GRCh38
rs751523943 Chr.16: 89583990-89583990 on Build GRCh38

View Full Product Details