Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 92353533-92353760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT3
Entrez Gene ID
120114
Gene Name
FAT atypical cadherin 3
Gene Aliases
CDHF15, CDHR10, hFat3
UniGene
Hs.98523
Gene Chromosome Location
Chr.11: 92224636-92896473 on Build GRCh38
Transcripts
NM_001008781.2 XM_017017178.1 XM_017017179.1 XM_017017180.1 XM_017017181.1 XM_017017182.1 XM_017017183.1 XM_017017184.1 XM_017017186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1561927 c.1546T>A p.L516M Chr.11: 92353658-92353658 on Build GRCh38
3782690 c.1461G>A p.V487V Chr.11: 92353573-92353573 on Build GRCh38
3782688 c.1461G>A p.V487V Chr.11: 92353573-92353573 on Build GRCh38
5106047 c.1593T>C p.T531T Chr.11: 92353705-92353705 on Build GRCh38
5788179 c.1487C>A p.A496D Chr.11: 92353599-92353599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032110961 Chr.11: 92353710-92353710 on Build GRCh38
rs771790688 Chr.11: 92353656-92353656 on Build GRCh38
rs1052725395 Chr.11: 92353672-92353672 on Build GRCh38
rs369038969 Chr.11: 92353575-92353575 on Build GRCh38
rs934380169 Chr.11: 92353671-92353671 on Build GRCh38
rs373479227 Chr.11: 92353582-92353582 on Build GRCh38
rs376399512 Chr.11: 92353583-92353583 on Build GRCh38
rs192455612 Chr.11: 92353646-92353646 on Build GRCh38
rs373120197 Chr.11: 92353590-92353590 on Build GRCh38
rs762947535 Chr.11: 92353560-92353560 on Build GRCh38
rs778023453 Chr.11: 92353645-92353645 on Build GRCh38
rs753240841 Chr.11: 92353628-92353628 on Build GRCh38
rs768595693 Chr.11: 92353694-92353694 on Build GRCh38
rs765868441 Chr.11: 92353619-92353619 on Build GRCh38
rs781507515 Chr.11: 92353655-92353655 on Build GRCh38
rs771837329 Chr.11: 92353721-92353721 on Build GRCh38
rs780252838 Chr.11: 92353675-92353675 on Build GRCh38
rs1019939580 Chr.11: 92353663-92353663 on Build GRCh38
rs757425400 Chr.11: 92353647-92353647 on Build GRCh38
rs377241528 Chr.11: 92353612-92353612 on Build GRCh38
rs774018033 Chr.11: 92353695-92353695 on Build GRCh38
rs758913947 Chr.11: 92353641-92353641 on Build GRCh38
rs559088371 Chr.11: 92353673-92353673 on Build GRCh38
rs139595720 Chr.11: 92353662-92353662 on Build GRCh38
rs774176716 Chr.11: 92353576-92353576 on Build GRCh38
rs1044177176 Chr.11: 92353703-92353703 on Build GRCh38
rs374543035 Chr.11: 92353708-92353708 on Build GRCh38
rs749235804 Chr.11: 92353688-92353688 on Build GRCh38

View Full Product Details