Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 115675965-115676238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAP43
Entrez Gene ID
2596
Gene Name
growth associated protein 43
Gene Aliases
B-50, PP46
UniGene
Hs.134974
Gene Chromosome Location
Chr.3: 115623304-115721487 on Build GRCh38
Transcripts
NM_001130064.1 NM_002045.3 XM_017006128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1149562 c.295G>A p.E99K Chr.3: 115676169-115676169 on Build GRCh38
5799013 c.103G>A p.A35T Chr.3: 115676085-115676085 on Build GRCh38
4686394 c.213A>C p.A71A Chr.3: 115676087-115676087 on Build GRCh38
3914609 c.49G>A p.D17N Chr.3: 115676031-115676031 on Build GRCh38
1149565 c.333C>G p.A111A Chr.3: 115676207-115676207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534171247 Chr.3: 115676181-115676181 on Build GRCh38
rs757884637 Chr.3: 115676020-115676020 on Build GRCh38
rs966472208 Chr.3: 115676152-115676152 on Build GRCh38
rs747086289 Chr.3: 115676179-115676179 on Build GRCh38
rs764745247 Chr.3: 115676089-115676089 on Build GRCh38
rs1029993074 Chr.3: 115676164-115676164 on Build GRCh38
rs759973547 Chr.3: 115676085-115676085 on Build GRCh38
rs139764193 Chr.3: 115676054-115676054 on Build GRCh38
rs762664283 Chr.3: 115676103-115676103 on Build GRCh38
rs763584753 Chr.3: 115676104-115676104 on Build GRCh38
rs750342541 Chr.3: 115676214-115676214 on Build GRCh38
rs781732711 Chr.3: 115676182-115676182 on Build GRCh38
rs767522147 Chr.3: 115676142-115676142 on Build GRCh38
rs912912456 Chr.3: 115676021-115676021 on Build GRCh38
rs368611099 Chr.3: 115676210-115676210 on Build GRCh38
rs953927987 Chr.3: 115676175-115676175 on Build GRCh38
rs747505118 Chr.3: 115676081-115676081 on Build GRCh38
rs762538944 Chr.3: 115676204-115676204 on Build GRCh38
rs1014993245 Chr.3: 115676118-115676118 on Build GRCh38
rs775179491 Chr.3: 115676090-115676090 on Build GRCh38
rs757246883 Chr.3: 115676031-115676031 on Build GRCh38
rs770193171 Chr.3: 115676199-115676199 on Build GRCh38
rs11557763 Chr.3: 115676051-115676051 on Build GRCh38
rs144973412 Chr.3: 115676005-115676005 on Build GRCh38
rs751087225 Chr.3: 115676110-115676110 on Build GRCh38
rs777937866 Chr.3: 115676166-115676166 on Build GRCh38
rs758751303 Chr.3: 115676159-115676159 on Build GRCh38
rs771573504 Chr.3: 115676082-115676082 on Build GRCh38
rs773812326 Chr.3: 115676208-115676208 on Build GRCh38
rs192134368 Chr.3: 115676052-115676052 on Build GRCh38
rs746404947 Chr.3: 115676185-115676185 on Build GRCh38
rs145057025 Chr.3: 115676084-115676084 on Build GRCh38
rs985218247 Chr.3: 115675993-115675993 on Build GRCh38
rs11557764 Chr.3: 115676058-115676058 on Build GRCh38
rs561268447 Chr.3: 115676050-115676050 on Build GRCh38
rs374365478 Chr.3: 115676048-115676048 on Build GRCh38
rs147441474 Chr.3: 115676030-115676030 on Build GRCh38
rs766987098 Chr.3: 115676213-115676213 on Build GRCh38
rs147136061 Chr.3: 115676114-115676114 on Build GRCh38
rs552993601 Chr.3: 115676207-115676207 on Build GRCh38
rs779752176 Chr.3: 115676155-115676155 on Build GRCh38
rs1032845517 Chr.3: 115676191-115676191 on Build GRCh38
rs763636093 Chr.3: 115676022-115676022 on Build GRCh38
rs201526194 Chr.3: 115676013-115676013 on Build GRCh38
rs780792860 Chr.3: 115676036-115676036 on Build GRCh38
rs113785130 Chr.3: 115676010-115676010 on Build GRCh38
rs201553710 Chr.3: 115676040-115676040 on Build GRCh38
rs370921978 Chr.3: 115676006-115676006 on Build GRCh38
rs6291 Chr.3: 115676157-115676157 on Build GRCh38
rs957182962 Chr.3: 115676196-115676196 on Build GRCh38
rs755917237 Chr.3: 115676151-115676151 on Build GRCh38
rs775779477 Chr.3: 115676202-115676202 on Build GRCh38
rs28399390 Chr.3: 115676150-115676150 on Build GRCh38

View Full Product Details