Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158684459-158684694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFDH
Entrez Gene ID
2110
Gene Name
electron transfer flavoprotein dehydrogenase
Gene Aliases
ETFQO, MADD
UniGene
Hs.155729
Gene Chromosome Location
Chr.4: 158672101-158708713 on Build GRCh38
Transcripts
NM_001281737.1 NM_001281738.1 NM_004453.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
310941 c.487G>C p.G163R Chr.4: 158684673-158684673 on Build GRCh38
6656644 c.445A>G p.I149V Chr.4: 158684631-158684631 on Build GRCh38
1052671 c.481C>A p.L161I Chr.4: 158684667-158684667 on Build GRCh38
4628271 c.429A>C p.T143T Chr.4: 158684615-158684615 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770638003 Chr.4: 158684566-158684567 on Build GRCh38
rs369257704 Chr.4: 158684553-158684553 on Build GRCh38
rs1019854055 Chr.4: 158684596-158684596 on Build GRCh38
rs1000355136 Chr.4: 158684484-158684484 on Build GRCh38
rs770555723 Chr.4: 158684582-158684582 on Build GRCh38
rs1007578889 Chr.4: 158684543-158684543 on Build GRCh38
rs779896449 Chr.4: 158684599-158684599 on Build GRCh38
rs751256504 Chr.4: 158684600-158684600 on Build GRCh38
rs999694563 Chr.4: 158684615-158684615 on Build GRCh38
rs758242572 Chr.4: 158684593-158684593 on Build GRCh38
rs773501580 Chr.4: 158684542-158684542 on Build GRCh38
rs549150456 Chr.4: 158684649-158684649 on Build GRCh38
rs780698954 Chr.4: 158684606-158684606 on Build GRCh38
rs759678812 Chr.4: 158684569-158684569 on Build GRCh38
rs995475699 Chr.4: 158684492-158684492 on Build GRCh38
rs878853082 Chr.4: 158684648-158684648 on Build GRCh38
rs753731596 Chr.4: 158684572-158684572 on Build GRCh38
rs766831541 Chr.4: 158684664-158684664 on Build GRCh38
rs754506098 Chr.4: 158684605-158684605 on Build GRCh38
rs146982178 Chr.4: 158684610-158684610 on Build GRCh38
rs771279534 Chr.4: 158684655-158684655 on Build GRCh38
rs764970272 Chr.4: 158684584-158684584 on Build GRCh38
rs767620024 Chr.4: 158684570-158684570 on Build GRCh38
rs373220891 Chr.4: 158684554-158684554 on Build GRCh38
rs774614480 Chr.4: 158684658-158684658 on Build GRCh38
rs749702132 Chr.4: 158684645-158684645 on Build GRCh38
rs773657159 Chr.4: 158684487-158684487 on Build GRCh38
rs1048353885 Chr.4: 158684498-158684498 on Build GRCh38
rs138149265 Chr.4: 158684627-158684627 on Build GRCh38
rs746043622 Chr.4: 158684666-158684666 on Build GRCh38
rs763492207 Chr.4: 158684607-158684607 on Build GRCh38
rs549069699 Chr.4: 158684488-158684488 on Build GRCh38
rs750189181 Chr.4: 158684591-158684591 on Build GRCh38
rs774524753 Chr.4: 158684567-158684567 on Build GRCh38
rs890064123 Chr.4: 158684524-158684524 on Build GRCh38

View Full Product Details