Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 88794421-88794652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf62
Entrez Gene ID
219557
Gene Name
chromosome 7 open reading frame 62
Gene Aliases
-
UniGene
Hs.112877
Gene Chromosome Location
Chr.7: 88794106-88797937 on Build GRCh38
Transcripts
NM_152706.3 XM_011515904.2 NM_181646.3

Gene Symbol
ZNF804B
Entrez Gene ID
219578
Gene Name
zinc finger protein 804B
Gene Aliases
-
UniGene
Hs.684410
Gene Chromosome Location
Chr.7: 88759368-89337057 on Build GRCh38
Transcripts
NM_152706.3 XM_011515904.2 NM_181646.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
747679 c.491C>T p.S164L Chr.7: 88794452-88794452 on Build GRCh38
5615890 c.475G>T p.D159Y Chr.7: 88794468-88794468 on Build GRCh38
3883142 c.355A>C p.K119Q Chr.7: 88794588-88794588 on Build GRCh38
271856 c.337G>T p.D113Y Chr.7: 88794606-88794606 on Build GRCh38
270948 c.382C>T p.Q128* Chr.7: 88794561-88794561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781359459 Chr.7: 88794574-88794574 on Build GRCh38
rs776842699 Chr.7: 88794464-88794464 on Build GRCh38
rs867402496 Chr.7: 88794556-88794556 on Build GRCh38
rs377269794 Chr.7: 88794469-88794469 on Build GRCh38
rs769803138 Chr.7: 88794577-88794577 on Build GRCh38
rs1054263128 Chr.7: 88794528-88794528 on Build GRCh38
rs747327078 Chr.7: 88794455-88794455 on Build GRCh38
rs372317539 Chr.7: 88794468-88794468 on Build GRCh38
rs756967541 Chr.7: 88794612-88794612 on Build GRCh38
rs775402208 Chr.7: 88794593-88794593 on Build GRCh38
rs763858589 Chr.7: 88794608-88794608 on Build GRCh38
rs776031300 Chr.7: 88794450-88794450 on Build GRCh38
rs780646965 Chr.7: 88794562-88794562 on Build GRCh38
rs745333051 Chr.7: 88794519-88794519 on Build GRCh38
rs758993492 Chr.7: 88794549-88794549 on Build GRCh38
rs143068564 Chr.7: 88794448-88794448 on Build GRCh38
rs756770295 Chr.7: 88794505-88794505 on Build GRCh38
rs377395892 Chr.7: 88794583-88794583 on Build GRCh38
rs764751270 Chr.7: 88794625-88794625 on Build GRCh38
rs758766386 Chr.7: 88794446-88794446 on Build GRCh38
rs2293583 Chr.7: 88794567-88794567 on Build GRCh38
rs752504765 Chr.7: 88794474-88794474 on Build GRCh38
rs201260308 Chr.7: 88794626-88794626 on Build GRCh38
rs146825913 Chr.7: 88794631-88794631 on Build GRCh38
rs767449422 Chr.7: 88794591-88794591 on Build GRCh38
rs758809917 Chr.7: 88794592-88794600 on Build GRCh38
rs753478549 Chr.7: 88794609-88794609 on Build GRCh38
rs200976271 Chr.7: 88794578-88794578 on Build GRCh38
rs966166733 Chr.7: 88794619-88794619 on Build GRCh38
rs369625195 Chr.7: 88794466-88794466 on Build GRCh38
rs34781240 Chr.7: 88794465-88794465 on Build GRCh38
rs997612987 Chr.7: 88794611-88794611 on Build GRCh38
rs533500155 Chr.7: 88794565-88794565 on Build GRCh38
rs755840841 Chr.7: 88794476-88794476 on Build GRCh38
rs749960309 Chr.7: 88794630-88794630 on Build GRCh38
rs201329404 Chr.7: 88794459-88794459 on Build GRCh38
rs747747087 Chr.7: 88794519-88794519 on Build GRCh38
rs369839861 Chr.7: 88794501-88794501 on Build GRCh38
rs143550892 Chr.7: 88794602-88794602 on Build GRCh38

View Full Product Details