Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3021811-3022084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTRT2
Entrez Gene ID
140625
Gene Name
actin related protein T2
Gene Aliases
ARPM2, ARPT2, Arp-T2, HARPM2
UniGene
Hs.236635
Gene Chromosome Location
Chr.1: 3021482-3022903 on Build GRCh38
Transcripts
NM_080431.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181750 c.167A>G p.Y56C Chr.1: 3021853-3021853 on Build GRCh38
2076418 c.397G>C p.V133L Chr.1: 3022083-3022083 on Build GRCh38
5713020 c.158_160delAGA p.K55delK Chr.1: 3021844-3021846 on Build GRCh38
6540014 c.176G>T p.G59V Chr.1: 3021862-3021862 on Build GRCh38
5733729 c.230G>A p.R77H Chr.1: 3021916-3021916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114122096 Chr.1: 3021860-3021860 on Build GRCh38
rs1043055524 Chr.1: 3021859-3021859 on Build GRCh38
rs748401902 Chr.1: 3021909-3021909 on Build GRCh38
rs35806103 Chr.1: 3022005-3022005 on Build GRCh38
rs778675360 Chr.1: 3021945-3021946 on Build GRCh38
rs756159916 Chr.1: 3021872-3021872 on Build GRCh38
rs776228150 Chr.1: 3021915-3021915 on Build GRCh38
rs373563032 Chr.1: 3022015-3022015 on Build GRCh38
rs761604182 Chr.1: 3022044-3022044 on Build GRCh38
rs755066337 Chr.1: 3021995-3021995 on Build GRCh38
rs144521527 Chr.1: 3021911-3021911 on Build GRCh38
rs1006784944 Chr.1: 3021905-3021905 on Build GRCh38
rs779098915 Chr.1: 3021835-3021835 on Build GRCh38
rs774096073 Chr.1: 3022041-3022041 on Build GRCh38
rs757797342 Chr.1: 3022013-3022013 on Build GRCh38
rs372637653 Chr.1: 3021899-3021899 on Build GRCh38
rs767477897 Chr.1: 3021984-3021984 on Build GRCh38
rs753755620 Chr.1: 3021855-3021855 on Build GRCh38
rs753937591 Chr.1: 3021878-3021878 on Build GRCh38
rs756036145 Chr.1: 3021839-3021840 on Build GRCh38
rs35037954 Chr.1: 3022016-3022016 on Build GRCh38
rs139603921 Chr.1: 3021864-3021864 on Build GRCh38
rs373435925 Chr.1: 3021903-3021903 on Build GRCh38
rs573881910 Chr.1: 3021858-3021858 on Build GRCh38
rs866545964 Chr.1: 3021861-3021861 on Build GRCh38
rs753027340 Chr.1: 3022004-3022004 on Build GRCh38
rs747558428 Chr.1: 3021912-3021912 on Build GRCh38
rs760247925 Chr.1: 3021856-3021856 on Build GRCh38
rs769262278 Chr.1: 3021933-3021933 on Build GRCh38
rs751477425 Chr.1: 3021967-3021967 on Build GRCh38
rs754826439 Chr.1: 3021865-3021882 on Build GRCh38
rs151321102 Chr.1: 3021916-3021916 on Build GRCh38
rs35209244 Chr.1: 3021831-3021831 on Build GRCh38
rs755154286 Chr.1: 3021883-3021883 on Build GRCh38
rs1055654275 Chr.1: 3021842-3021842 on Build GRCh38
rs778249092 Chr.1: 3022008-3022008 on Build GRCh38
rs749018428 Chr.1: 3022038-3022038 on Build GRCh38
rs1020171023 Chr.1: 3021929-3021929 on Build GRCh38
rs747399093 Chr.1: 3022011-3022011 on Build GRCh38
rs772395277 Chr.1: 3021841-3021841 on Build GRCh38
rs559954352 Chr.1: 3021983-3021983 on Build GRCh38
rs768220031 Chr.1: 3022040-3022040 on Build GRCh38
rs778581059 Chr.1: 3022046-3022046 on Build GRCh38
rs200744486 Chr.1: 3022033-3022033 on Build GRCh38
rs370720529 Chr.1: 3021990-3021990 on Build GRCh38
rs140563911 Chr.1: 3021941-3021941 on Build GRCh38
rs558364798 Chr.1: 3021996-3021996 on Build GRCh38
rs141084585 Chr.1: 3021943-3021943 on Build GRCh38
rs34872045 Chr.1: 3021942-3021942 on Build GRCh38
rs4013154 Chr.1: 3021844-3021846 on Build GRCh38
rs201756456 Chr.1: 3022045-3022045 on Build GRCh38
rs1018621832 Chr.1: 3021907-3021907 on Build GRCh38
rs137950147 Chr.1: 3021852-3021852 on Build GRCh38
rs748578742 Chr.1: 3021838-3021838 on Build GRCh38
rs770953749 Chr.1: 3021863-3021863 on Build GRCh38
rs201580305 Chr.1: 3021846-3021846 on Build GRCh38
rs780157065 Chr.1: 3021875-3021875 on Build GRCh38

View Full Product Details