Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41663584-41663823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK1
Entrez Gene ID
286
Gene Name
ankyrin 1
Gene Aliases
ANK, SPH1, SPH2
UniGene
Hs.654438
Gene Chromosome Location
Chr.8: 41653225-41896762 on Build GRCh38
Transcripts
NM_000037.3 NM_001142445.1 NM_001142446.1 NM_020475.2 NM_020476.2 NM_020477.2 NM_020478.4 NM_020480.4 XM_005273476.4 XM_011544490.2 XM_011544491.2 XM_011544494.2 XM_011544495.2 XM_011544496.2 XM_011544500.2 XM_011544501.2 XM_011544502.2 XM_011544503.2 XM_011544504.2 XM_011544505.2 XM_017013319.1 XM_017013320.1 XM_017013321.1 XM_017013322.1 XM_017013323.1 XM_017013324.1 XM_017013325.1 XM_017013326.1 XM_017013327.1 XM_017013328.1 XM_017013329.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5932196 c.262G>A p.E88K Chr.8: 41663700-41663700 on Build GRCh38
5000139 c.5519G>A p.G1840E Chr.8: 41663741-41663741 on Build GRCh38
7303277 c.5518-4C>T p.? Chr.8: 41663746-41663746 on Build GRCh38
6599730 c.5447C>T p.T1816M Chr.8: 41663690-41663690 on Build GRCh38
6599729 c.5570C>T p.T1857M Chr.8: 41663690-41663690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953540786 Chr.8: 41663663-41663663 on Build GRCh38
rs984851920 Chr.8: 41663698-41663698 on Build GRCh38
rs767694632 Chr.8: 41663775-41663775 on Build GRCh38
rs924618480 Chr.8: 41663796-41663796 on Build GRCh38
rs990648239 Chr.8: 41663725-41663725 on Build GRCh38
rs767639445 Chr.8: 41663629-41663629 on Build GRCh38
rs949308336 Chr.8: 41663744-41663744 on Build GRCh38
rs751709273 Chr.8: 41663625-41663625 on Build GRCh38
rs759817762 Chr.8: 41663767-41663767 on Build GRCh38
rs752870311 Chr.8: 41663777-41663777 on Build GRCh38
rs747582386 Chr.8: 41663710-41663710 on Build GRCh38
rs780824103 Chr.8: 41663709-41663709 on Build GRCh38
rs750253055 Chr.8: 41663672-41663672 on Build GRCh38
rs763307966 Chr.8: 41663758-41663758 on Build GRCh38
rs368285911 Chr.8: 41663674-41663674 on Build GRCh38
rs200151645 Chr.8: 41663761-41663761 on Build GRCh38
rs1042285116 Chr.8: 41663770-41663770 on Build GRCh38
rs778693874 Chr.8: 41663637-41663637 on Build GRCh38
rs146637140 Chr.8: 41663689-41663689 on Build GRCh38
rs528332130 Chr.8: 41663679-41663679 on Build GRCh38
rs773545929 Chr.8: 41663740-41663740 on Build GRCh38
rs774682195 Chr.8: 41663764-41663764 on Build GRCh38
rs757257850 Chr.8: 41663635-41663635 on Build GRCh38
rs778838145 Chr.8: 41663690-41663690 on Build GRCh38
rs752714181 Chr.8: 41663631-41663631 on Build GRCh38
rs770917173 Chr.8: 41663658-41663658 on Build GRCh38
rs770385311 Chr.8: 41663718-41663718 on Build GRCh38
rs771224017 Chr.8: 41663763-41663763 on Build GRCh38
rs766615613 Chr.8: 41663618-41663618 on Build GRCh38
rs917718228 Chr.8: 41663735-41663735 on Build GRCh38
rs201031196 Chr.8: 41663613-41663613 on Build GRCh38
rs755062097 Chr.8: 41663626-41663626 on Build GRCh38
rs371526642 Chr.8: 41663617-41663617 on Build GRCh38

View Full Product Details