Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231256475-231256728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC9
Entrez Gene ID
80210
Gene Name
armadillo repeat containing 9
Gene Aliases
ARM, KU-MEL-1, NS21
UniGene
Hs.162411
Gene Chromosome Location
Chr.2: 231198546-231394991 on Build GRCh38
Transcripts
NM_001271466.2 NM_001291656.1 NM_025139.5 XM_011511905.1 XM_011511906.1 XM_011511907.1 XM_011511908.2 XM_011511909.2 XM_011511910.1 XM_011511911.1 XM_011511912.1 XM_011511913.2 XM_011511914.2 XM_011511915.2 XM_011511916.2 XM_017005018.1 XM_017005019.1 XM_017005020.1 XM_017005021.1 XM_017005022.1 XM_017005023.1 XM_017005024.1 XM_017005025.1 XM_017005026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5917531 c.902C>T p.S301F Chr.2: 231256608-231256608 on Build GRCh38
1641862 c.880-8_880-7insC p.? Chr.2: 231256578-231256579 on Build GRCh38
4663420 c.912C>T p.P304P Chr.2: 231256618-231256618 on Build GRCh38
3838832 c.880-1G>A p.? Chr.2: 231256585-231256585 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs539071524 Chr.2: 231256682-231256682 on Build GRCh38
rs779641672 Chr.2: 231256593-231256593 on Build GRCh38
rs74816123 Chr.2: 231256694-231256694 on Build GRCh38
rs777521211 Chr.2: 231256636-231256636 on Build GRCh38
rs565930480 Chr.2: 231256675-231256675 on Build GRCh38
rs755801320 Chr.2: 231256588-231256588 on Build GRCh38
rs1046156279 Chr.2: 231256655-231256655 on Build GRCh38
rs778488064 Chr.2: 231256604-231256604 on Build GRCh38
rs764248634 Chr.2: 231256576-231256576 on Build GRCh38
rs750275696 Chr.2: 231256526-231256526 on Build GRCh38
rs750869801 Chr.2: 231256583-231256583 on Build GRCh38
rs771634496 Chr.2: 231256650-231256652 on Build GRCh38
rs906264863 Chr.2: 231256673-231256673 on Build GRCh38
rs775607078 Chr.2: 231256535-231256535 on Build GRCh38
rs767945191 Chr.2: 231256582-231256582 on Build GRCh38
rs775010792 Chr.2: 231256651-231256653 on Build GRCh38
rs776663852 Chr.2: 231256642-231256642 on Build GRCh38
rs753378948 Chr.2: 231256597-231256597 on Build GRCh38
rs535773953 Chr.2: 231256578-231256578 on Build GRCh38
rs547395009 Chr.2: 231256641-231256641 on Build GRCh38
rs900805098 Chr.2: 231256536-231256536 on Build GRCh38
rs1053897537 Chr.2: 231256557-231256557 on Build GRCh38
rs772015381 Chr.2: 231256630-231256630 on Build GRCh38
rs754385274 Chr.2: 231256601-231256601 on Build GRCh38
rs748033626 Chr.2: 231256619-231256619 on Build GRCh38
rs775630320 Chr.2: 231256653-231256653 on Build GRCh38
rs939912615 Chr.2: 231256510-231256510 on Build GRCh38
rs367798821 Chr.2: 231256543-231256543 on Build GRCh38
rs936225525 Chr.2: 231256550-231256550 on Build GRCh38
rs540126371 Chr.2: 231256579-231256579 on Build GRCh38
rs1002013879 Chr.2: 231256674-231256674 on Build GRCh38
rs370482100 Chr.2: 231256517-231256517 on Build GRCh38
rs568243247 Chr.2: 231256530-231256530 on Build GRCh38
rs768579908 Chr.2: 231256667-231256667 on Build GRCh38
rs750727367 Chr.2: 231256663-231256663 on Build GRCh38
rs996922516 Chr.2: 231256618-231256618 on Build GRCh38
rs112171686 Chr.2: 231256529-231256529 on Build GRCh38

View Full Product Details