Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117291631-117291892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACE1
Entrez Gene ID
23621
Gene Name
beta-secretase 1
Gene Aliases
ASP2, BACE, HSPC104
UniGene
Hs.504003
Gene Chromosome Location
Chr.11: 117285686-117316256 on Build GRCh38
Transcripts
NM_001207048.1 NM_001207049.1 NM_012104.4 NM_138971.3 NM_138972.3 NM_138973.3 NR_037803.2

Gene Symbol
BACE1-AS
Entrez Gene ID
100379571
Gene Name
BACE1 antisense RNA
Gene Aliases
BACE1-AS1, BACE1AS, NCRNA00177
UniGene
Hs.736305
Gene Chromosome Location
Chr.11: 117291346-117292170 on Build GRCh38
Transcripts
NM_001207048.1 NM_001207049.1 NM_012104.4 NM_138971.3 NM_138972.3 NM_138973.3 NR_037803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5349619 c.847T>C p.Y283H Chr.11: 117291807-117291807 on Build GRCh38
4018146 c.942+2T>A p.? Chr.11: 117291710-117291710 on Build GRCh38
2104604 c.942C>T p.S314S Chr.11: 117291712-117291712 on Build GRCh38
5837879 c.872G>A p.G291D Chr.11: 117291782-117291782 on Build GRCh38
4969703 c.887G>A p.R296H Chr.11: 117291767-117291767 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780202200 Chr.11: 117291827-117291827 on Build GRCh38
rs747913030 Chr.11: 117291707-117291707 on Build GRCh38
rs997611644 Chr.11: 117291759-117291759 on Build GRCh38
rs779087846 Chr.11: 117291682-117291682 on Build GRCh38
rs775866522 Chr.11: 117291751-117291751 on Build GRCh38
rs61903702 Chr.11: 117291772-117291772 on Build GRCh38
rs1015098316 Chr.11: 117291684-117291684 on Build GRCh38
rs768494471 Chr.11: 117291697-117291697 on Build GRCh38
rs577986178 Chr.11: 117291656-117291656 on Build GRCh38
rs370895260 Chr.11: 117291864-117291864 on Build GRCh38
rs767280509 Chr.11: 117291738-117291738 on Build GRCh38
rs776542380 Chr.11: 117291826-117291827 on Build GRCh38
rs367712645 Chr.11: 117291844-117291844 on Build GRCh38
rs147542312 Chr.11: 117291711-117291711 on Build GRCh38
rs777739218 Chr.11: 117291838-117291838 on Build GRCh38
rs573665061 Chr.11: 117291689-117291689 on Build GRCh38
rs747973911 Chr.11: 117291829-117291829 on Build GRCh38
rs758654476 Chr.11: 117291690-117291690 on Build GRCh38
rs757786235 Chr.11: 117291781-117291781 on Build GRCh38
rs774018648 Chr.11: 117291729-117291729 on Build GRCh38
rs759223338 Chr.11: 117291736-117291736 on Build GRCh38
rs769761591 Chr.11: 117291709-117291709 on Build GRCh38
rs761525173 Chr.11: 117291794-117291794 on Build GRCh38
rs917696892 Chr.11: 117291694-117291694 on Build GRCh38
rs746782706 Chr.11: 117291684-117291685 on Build GRCh38
rs753368313 Chr.11: 117291756-117291756 on Build GRCh38
rs542748548 Chr.11: 117291719-117291719 on Build GRCh38
rs755898660 Chr.11: 117291834-117291834 on Build GRCh38
rs540255037 Chr.11: 117291659-117291659 on Build GRCh38
rs780900259 Chr.11: 117291697-117291697 on Build GRCh38
rs150090942 Chr.11: 117291665-117291665 on Build GRCh38
rs562442754 Chr.11: 117291805-117291805 on Build GRCh38
rs1048617899 Chr.11: 117291658-117291658 on Build GRCh38
rs773929696 Chr.11: 117291849-117291849 on Build GRCh38
rs908883505 Chr.11: 117291874-117291874 on Build GRCh38
rs749128675 Chr.11: 117291839-117291839 on Build GRCh38
rs764521142 Chr.11: 117291664-117291664 on Build GRCh38
rs765262339 Chr.11: 117291782-117291782 on Build GRCh38
rs748797299 Chr.11: 117291712-117291712 on Build GRCh38
rs200390419 Chr.11: 117291685-117291685 on Build GRCh38
rs761002162 Chr.11: 117291757-117291757 on Build GRCh38
rs556962526 Chr.11: 117291718-117291718 on Build GRCh38
rs754352311 Chr.11: 117291771-117291771 on Build GRCh38
rs756577849 Chr.11: 117291662-117291662 on Build GRCh38
rs531164090 Chr.11: 117291859-117291859 on Build GRCh38
rs758561454 Chr.11: 117291826-117291826 on Build GRCh38

View Full Product Details