Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20785927-20786162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
UniGene
Hs.706754
Gene Chromosome Location
Chr.16: 20763990-20797157 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Gene Symbol
ERI2
Entrez Gene ID
112479
Gene Name
ERI1 exoribonuclease family member 2
Gene Aliases
EXOD1, ZGRF5
UniGene
Hs.248437
Gene Chromosome Location
Chr.16: 20780193-20806473 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5730287 c.1171A>G p.M391V Chr.16: 20786105-20786105 on Build GRCh38
1708871 c.1168G>A p.G390R Chr.16: 20786102-20786102 on Build GRCh38
3507066 c.1173G>A p.M391I Chr.16: 20786107-20786107 on Build GRCh38
4659056 c.1202_1204delCTT p.S402delS Chr.16: 20786136-20786138 on Build GRCh38
4659057 c.1202_1204delCTT p.S402delS Chr.16: 20786136-20786138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757329851 Chr.16: 20786114-20786114 on Build GRCh38
rs375425139 Chr.16: 20786060-20786060 on Build GRCh38
rs772817635 Chr.16: 20786032-20786032 on Build GRCh38
rs1033509057 Chr.16: 20786068-20786068 on Build GRCh38
rs752632741 Chr.16: 20786113-20786113 on Build GRCh38
rs755856152 Chr.16: 20786042-20786046 on Build GRCh38
rs758636757 Chr.16: 20786124-20786124 on Build GRCh38
rs762548951 Chr.16: 20786054-20786054 on Build GRCh38
rs777074557 Chr.16: 20786028-20786028 on Build GRCh38
rs767483367 Chr.16: 20786096-20786096 on Build GRCh38
rs765370178 Chr.16: 20786117-20786117 on Build GRCh38
rs896004078 Chr.16: 20786013-20786013 on Build GRCh38
rs760002342 Chr.16: 20786069-20786069 on Build GRCh38
rs780432331 Chr.16: 20786136-20786136 on Build GRCh38
rs750587830 Chr.16: 20786061-20786063 on Build GRCh38
rs759253257 Chr.16: 20786088-20786088 on Build GRCh38
rs897404223 Chr.16: 20786058-20786058 on Build GRCh38
rs201784927 Chr.16: 20786031-20786031 on Build GRCh38
rs72778664 Chr.16: 20786038-20786038 on Build GRCh38
rs764006483 Chr.16: 20786053-20786056 on Build GRCh38
rs1034448618 Chr.16: 20785980-20785980 on Build GRCh38
rs978820171 Chr.16: 20786047-20786047 on Build GRCh38
rs747683146 Chr.16: 20786029-20786029 on Build GRCh38
rs368608072 Chr.16: 20786120-20786120 on Build GRCh38
rs766038257 Chr.16: 20786076-20786076 on Build GRCh38
rs267604444 Chr.16: 20786107-20786107 on Build GRCh38
rs774073648 Chr.16: 20786082-20786082 on Build GRCh38

View Full Product Details