Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17543271-17543536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM129C
Entrez Gene ID
199786
Gene Name
family with sequence similarity 129 member C
Gene Aliases
BCNP1
UniGene
Hs.434133
Gene Chromosome Location
Chr.19: 17523301-17556755 on Build GRCh38
Transcripts
NM_001098524.1 NM_001321826.1 NM_001321827.1 NM_001321828.1 NM_173544.4 XM_005259813.3 XM_005259814.3 XM_011527781.2 XM_011527786.2 XM_011527787.2 XM_011527789.1 XM_017026453.1 XM_017026454.1 XM_017026455.1 XM_017026456.1 XM_017026457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7228891 c.1479G>A p.T493T Chr.19: 17543373-17543373 on Build GRCh38
6568130 c.1471C>T p.L491L Chr.19: 17543365-17543365 on Build GRCh38
3286302 c.1489T>C p.C497R Chr.19: 17543383-17543383 on Build GRCh38
6568132 c.1471C>T p.L491L Chr.19: 17543365-17543365 on Build GRCh38
6238127 c.1508G>A p.R503K Chr.19: 17543402-17543402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370587823 Chr.19: 17543505-17543505 on Build GRCh38
rs910301558 Chr.19: 17543491-17543491 on Build GRCh38
rs747029118 Chr.19: 17543423-17543423 on Build GRCh38
rs756361874 Chr.19: 17543503-17543503 on Build GRCh38
rs755327626 Chr.19: 17543408-17543408 on Build GRCh38
rs777645998 Chr.19: 17543506-17543506 on Build GRCh38
rs749046878 Chr.19: 17543363-17543363 on Build GRCh38
rs760303982 Chr.19: 17543395-17543395 on Build GRCh38
rs376170078 Chr.19: 17543297-17543297 on Build GRCh38
rs45532635 Chr.19: 17543372-17543372 on Build GRCh38
rs140048862 Chr.19: 17543383-17543383 on Build GRCh38
rs771587440 Chr.19: 17543500-17543500 on Build GRCh38
rs758503031 Chr.19: 17543332-17543332 on Build GRCh38
rs767040743 Chr.19: 17543296-17543296 on Build GRCh38
rs760108074 Chr.19: 17543457-17543457 on Build GRCh38
rs978298686 Chr.19: 17543426-17543426 on Build GRCh38
rs765996075 Chr.19: 17543397-17543397 on Build GRCh38
rs375915215 Chr.19: 17543450-17543450 on Build GRCh38
rs901752707 Chr.19: 17543300-17543300 on Build GRCh38
rs1033755750 Chr.19: 17543386-17543386 on Build GRCh38
rs748632827 Chr.19: 17543331-17543331 on Build GRCh38
rs753435874 Chr.19: 17543398-17543398 on Build GRCh38
rs139360897 Chr.19: 17543369-17543369 on Build GRCh38
rs770378293 Chr.19: 17543458-17543458 on Build GRCh38
rs763851677 Chr.19: 17543474-17543477 on Build GRCh38
rs764036830 Chr.19: 17543291-17543291 on Build GRCh38
rs772051831 Chr.19: 17543453-17543453 on Build GRCh38
rs778752017 Chr.19: 17543504-17543504 on Build GRCh38
rs755308299 Chr.19: 17543328-17543328 on Build GRCh38
rs761275168 Chr.19: 17543295-17543295 on Build GRCh38
rs746102574 Chr.19: 17543433-17543433 on Build GRCh38
rs777165201 Chr.19: 17543416-17543416 on Build GRCh38
rs750631966 Chr.19: 17543501-17543501 on Build GRCh38
rs776296067 Chr.19: 17543460-17543460 on Build GRCh38
rs753171590 Chr.19: 17543468-17543468 on Build GRCh38
rs755799945 Chr.19: 17543304-17543304 on Build GRCh38
rs765604812 Chr.19: 17543465-17543465 on Build GRCh38
rs373733258 Chr.19: 17543422-17543422 on Build GRCh38
rs752145381 Chr.19: 17543487-17543487 on Build GRCh38
rs747231201 Chr.19: 17543350-17543350 on Build GRCh38
rs768479344 Chr.19: 17543444-17543444 on Build GRCh38
rs773145349 Chr.19: 17543455-17543455 on Build GRCh38
rs757315091 Chr.19: 17543488-17543488 on Build GRCh38
rs750474277 Chr.19: 17543370-17543370 on Build GRCh38
rs76814516 Chr.19: 17543373-17543373 on Build GRCh38
rs766721817 Chr.19: 17543319-17543319 on Build GRCh38
rs138987050 Chr.19: 17543424-17543424 on Build GRCh38
rs753232898 Chr.19: 17543417-17543417 on Build GRCh38
rs779409000 Chr.19: 17543329-17543329 on Build GRCh38
rs746970135 Chr.19: 17543507-17543507 on Build GRCh38
rs555880628 Chr.19: 17543479-17543479 on Build GRCh38
rs888799687 Chr.19: 17543420-17543420 on Build GRCh38
rs374603681 Chr.19: 17543339-17543339 on Build GRCh38
rs936791404 Chr.19: 17543451-17543451 on Build GRCh38
rs763424298 Chr.19: 17543470-17543470 on Build GRCh38
rs771337988 Chr.19: 17543361-17543361 on Build GRCh38
rs762645388 Chr.19: 17543464-17543469 on Build GRCh38
rs200100942 Chr.19: 17543425-17543425 on Build GRCh38
rs759160481 Chr.19: 17543463-17543463 on Build GRCh38
rs369494726 Chr.19: 17543411-17543411 on Build GRCh38
rs550418533 Chr.19: 17543327-17543327 on Build GRCh38

View Full Product Details