Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 54164375-54164644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP24A1
Entrez Gene ID
1591
Gene Name
cytochrome P450 family 24 subfamily A member 1
Gene Aliases
CP24, CYP24, HCAI, HCINF1, P450-CC24
UniGene
Hs.89663
Gene Chromosome Location
Chr.20: 54145731-54173985 on Build GRCh38
Transcripts
NM_000782.4 NM_001128915.1 XM_005260304.4 XM_017027691.1 XM_017027692.1 XM_017027693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1412575 c.804G>T p.W268C Chr.20: 54164492-54164492 on Build GRCh38
3379090 c.806A>G p.Q269R Chr.20: 54164490-54164490 on Build GRCh38
5815408 c.816T>A p.T272T Chr.20: 54164480-54164480 on Build GRCh38
5952849 c.756G>C p.M252I Chr.20: 54164540-54164540 on Build GRCh38
6093880 c.831C>A p.T277T Chr.20: 54164465-54164465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952668735 Chr.20: 54164411-54164411 on Build GRCh38
rs777867868 Chr.20: 54164581-54164581 on Build GRCh38
rs373902459 Chr.20: 54164520-54164520 on Build GRCh38
rs781564921 Chr.20: 54164486-54164486 on Build GRCh38
rs373292619 Chr.20: 54164433-54164433 on Build GRCh38
rs763760614 Chr.20: 54164446-54164446 on Build GRCh38
rs368898455 Chr.20: 54164465-54164465 on Build GRCh38
rs577760976 Chr.20: 54164412-54164412 on Build GRCh38
rs751888651 Chr.20: 54164472-54164472 on Build GRCh38
rs375874703 Chr.20: 54164439-54164439 on Build GRCh38
rs1046447948 Chr.20: 54164616-54164616 on Build GRCh38
rs114930663 Chr.20: 54164561-54164561 on Build GRCh38
rs868568479 Chr.20: 54164512-54164512 on Build GRCh38
rs925534555 Chr.20: 54164547-54164547 on Build GRCh38
rs776333239 Chr.20: 54164515-54164515 on Build GRCh38
rs753887561 Chr.20: 54164582-54164582 on Build GRCh38
rs773374239 Chr.20: 54164405-54164405 on Build GRCh38
rs16999131 Chr.20: 54164553-54164553 on Build GRCh38
rs746821511 Chr.20: 54164508-54164508 on Build GRCh38
rs760886293 Chr.20: 54164404-54164405 on Build GRCh38
rs753016494 Chr.20: 54164570-54164570 on Build GRCh38
rs771949062 Chr.20: 54164434-54164434 on Build GRCh38
rs747308046 Chr.20: 54164405-54164405 on Build GRCh38
rs190860407 Chr.20: 54164522-54164522 on Build GRCh38
rs776062343 Chr.20: 54164450-54164450 on Build GRCh38
rs771933784 Chr.20: 54164497-54164497 on Build GRCh38
rs770921201 Chr.20: 54164420-54164420 on Build GRCh38
rs760337236 Chr.20: 54164441-54164441 on Build GRCh38
rs182327351 Chr.20: 54164583-54164583 on Build GRCh38
rs755190328 Chr.20: 54164482-54164482 on Build GRCh38
rs760041455 Chr.20: 54164558-54164558 on Build GRCh38
rs6097817 Chr.20: 54164438-54164438 on Build GRCh38
rs202109000 Chr.20: 54164525-54164525 on Build GRCh38
rs756319778 Chr.20: 54164489-54164489 on Build GRCh38
rs559914563 Chr.20: 54164428-54164428 on Build GRCh38
rs186004232 Chr.20: 54164404-54164404 on Build GRCh38
rs528653327 Chr.20: 54164456-54164456 on Build GRCh38
rs745856178 Chr.20: 54164494-54164494 on Build GRCh38
rs377533549 Chr.20: 54164543-54164543 on Build GRCh38
rs552082262 Chr.20: 54164467-54164467 on Build GRCh38
rs779851763 Chr.20: 54164501-54164501 on Build GRCh38
rs756229861 Chr.20: 54164575-54164575 on Build GRCh38
rs773564044 Chr.20: 54164537-54164537 on Build GRCh38
rs778693884 Chr.20: 54164492-54164492 on Build GRCh38
rs779721022 Chr.20: 54164521-54164521 on Build GRCh38
rs370079250 Chr.20: 54164402-54164402 on Build GRCh38
rs6068816 Chr.20: 54164552-54164552 on Build GRCh38
rs781368642 Chr.20: 54164417-54164417 on Build GRCh38
rs895309596 Chr.20: 54164572-54164572 on Build GRCh38
rs761318027 Chr.20: 54164454-54164454 on Build GRCh38
rs199934501 Chr.20: 54164513-54164513 on Build GRCh38

View Full Product Details