Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 14602134-14602389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM105A
Entrez Gene ID
54491
Gene Name
family with sequence similarity 105 member A
Gene Aliases
NET20
UniGene
Hs.155085
Gene Chromosome Location
Chr.5: 14581782-14616180 on Build GRCh38
Transcripts
NM_019018.2 XM_011514056.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5756591 c.367T>C p.W123R Chr.5: 14602201-14602201 on Build GRCh38
6568396 c.460T>A p.S154T Chr.5: 14602294-14602294 on Build GRCh38
1695468 c.466C>T p.P156S Chr.5: 14602300-14602300 on Build GRCh38
4840191 c.382A>G p.K128E Chr.5: 14602216-14602216 on Build GRCh38
3612041 c.402G>A p.R134R Chr.5: 14602236-14602236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749409435 Chr.5: 14602232-14602232 on Build GRCh38
rs778016855 Chr.5: 14602324-14602324 on Build GRCh38
rs773276642 Chr.5: 14602226-14602226 on Build GRCh38
rs755313253 Chr.5: 14602305-14602305 on Build GRCh38
rs747354708 Chr.5: 14602204-14602204 on Build GRCh38
rs891235736 Chr.5: 14602225-14602225 on Build GRCh38
rs574606640 Chr.5: 14602361-14602361 on Build GRCh38
rs767217886 Chr.5: 14602261-14602261 on Build GRCh38
rs200853586 Chr.5: 14602303-14602303 on Build GRCh38
rs145808843 Chr.5: 14602323-14602323 on Build GRCh38
rs371184498 Chr.5: 14602169-14602169 on Build GRCh38
rs1011146465 Chr.5: 14602214-14602214 on Build GRCh38
rs374030566 Chr.5: 14602300-14602300 on Build GRCh38
rs116666867 Chr.5: 14602294-14602294 on Build GRCh38
rs745782103 Chr.5: 14602300-14602300 on Build GRCh38
rs897038543 Chr.5: 14602177-14602177 on Build GRCh38
rs748615470 Chr.5: 14602306-14602306 on Build GRCh38
rs749316313 Chr.5: 14602342-14602342 on Build GRCh38
rs114630989 Chr.5: 14602284-14602284 on Build GRCh38
rs773657350 Chr.5: 14602336-14602336 on Build GRCh38
rs756853225 Chr.5: 14602279-14602279 on Build GRCh38
rs748332389 Chr.5: 14602213-14602213 on Build GRCh38
rs774499872 Chr.5: 14602296-14602296 on Build GRCh38
rs116207901 Chr.5: 14602262-14602262 on Build GRCh38
rs759298079 Chr.5: 14602165-14602165 on Build GRCh38
rs752319914 Chr.5: 14602173-14602173 on Build GRCh38
rs367770838 Chr.5: 14602231-14602231 on Build GRCh38
rs753453244 Chr.5: 14602188-14602188 on Build GRCh38
rs113242974 Chr.5: 14602190-14602190 on Build GRCh38
rs770831747 Chr.5: 14602242-14602242 on Build GRCh38
rs199720349 Chr.5: 14602270-14602270 on Build GRCh38
rs114672257 Chr.5: 14602249-14602249 on Build GRCh38
rs768937348 Chr.5: 14602205-14602205 on Build GRCh38
rs959082569 Chr.5: 14602289-14602289 on Build GRCh38
rs1050857858 Chr.5: 14602211-14602211 on Build GRCh38
rs199518529 Chr.5: 14602307-14602307 on Build GRCh38
rs770061268 Chr.5: 14602218-14602218 on Build GRCh38
rs763902840 Chr.5: 14602263-14602263 on Build GRCh38
rs767327303 Chr.5: 14602342-14602342 on Build GRCh38
rs777433281 Chr.5: 14602180-14602180 on Build GRCh38
rs758946237 Chr.5: 14602192-14602192 on Build GRCh38
rs780757565 Chr.5: 14602197-14602197 on Build GRCh38
rs755580099 Chr.5: 14602178-14602178 on Build GRCh38
rs144733724 Chr.5: 14602210-14602210 on Build GRCh38

View Full Product Details