Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47975814-47976051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP3
Entrez Gene ID
80279
Gene Name
CDK5 regulatory subunit associated protein 3
Gene Aliases
C53, HSF-27, IC53, LZAP, MST016, OK/SW-cl.114, PP1553
UniGene
Hs.20157
Gene Chromosome Location
Chr.17: 47967904-47981786 on Build GRCh38
Transcripts
NM_001278197.1 NM_001278198.1 NM_001278216.1 NM_001278217.1 NM_176096.2 XM_011525297.1 XM_017025164.1 XM_017025165.1 XM_017025166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5123016 c.754C>T p.R252* Chr.17: 47975969-47975969 on Build GRCh38
3387991 c.737A>C p.E246A Chr.17: 47975952-47975952 on Build GRCh38
5722637 c.784G>A p.V262M Chr.17: 47975999-47975999 on Build GRCh38
980572 c.753A>G p.E251E Chr.17: 47975968-47975968 on Build GRCh38
3712355 c.687C>T p.F229F Chr.17: 47975902-47975902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550711152 Chr.17: 47975910-47975910 on Build GRCh38
rs772953445 Chr.17: 47975969-47975969 on Build GRCh38
rs746686583 Chr.17: 47975970-47975970 on Build GRCh38
rs767898149 Chr.17: 47975927-47975927 on Build GRCh38
rs751914618 Chr.17: 47975862-47975862 on Build GRCh38
rs932272625 Chr.17: 47975960-47975960 on Build GRCh38
rs192721406 Chr.17: 47975998-47975998 on Build GRCh38
rs755014626 Chr.17: 47975962-47975962 on Build GRCh38
rs376737457 Chr.17: 47975932-47975932 on Build GRCh38
rs61753863 Chr.17: 47975897-47975897 on Build GRCh38
rs1019567616 Chr.17: 47975853-47975853 on Build GRCh38
rs763529107 Chr.17: 47975923-47975923 on Build GRCh38
rs770433684 Chr.17: 47975976-47975976 on Build GRCh38
rs938912587 Chr.17: 47975986-47975986 on Build GRCh38
rs375039682 Chr.17: 47976007-47976007 on Build GRCh38
rs775955130 Chr.17: 47975919-47975919 on Build GRCh38
rs764441122 Chr.17: 47975924-47975924 on Build GRCh38
rs762304936 Chr.17: 47975990-47975990 on Build GRCh38
rs781558811 Chr.17: 47975863-47975863 on Build GRCh38
rs752664351 Chr.17: 47976025-47976025 on Build GRCh38
rs575641521 Chr.17: 47975903-47975903 on Build GRCh38
rs757680785 Chr.17: 47975926-47975926 on Build GRCh38
rs199867075 Chr.17: 47976013-47976013 on Build GRCh38
rs779893428 Chr.17: 47975891-47975891 on Build GRCh38
rs780202262 Chr.17: 47975956-47975956 on Build GRCh38
rs769180917 Chr.17: 47975856-47975866 on Build GRCh38
rs965667352 Chr.17: 47975860-47975860 on Build GRCh38
rs200409765 Chr.17: 47975837-47975837 on Build GRCh38
rs1050645462 Chr.17: 47975973-47975973 on Build GRCh38
rs778865181 Chr.17: 47976023-47976023 on Build GRCh38
rs747988734 Chr.17: 47975966-47975966 on Build GRCh38
rs1054556333 Chr.17: 47975938-47975938 on Build GRCh38
rs774205338 Chr.17: 47975902-47975902 on Build GRCh38
rs113940013 Chr.17: 47976014-47976014 on Build GRCh38
rs766509639 Chr.17: 47976012-47976012 on Build GRCh38
rs776208156 Chr.17: 47975980-47975980 on Build GRCh38
rs374509909 Chr.17: 47975935-47975935 on Build GRCh38
rs377690706 Chr.17: 47975958-47975958 on Build GRCh38
rs752251451 Chr.17: 47975993-47975993 on Build GRCh38
rs1028129510 Chr.17: 47976022-47976022 on Build GRCh38
rs61752524 Chr.17: 47975925-47975925 on Build GRCh38
rs199625816 Chr.17: 47975912-47975912 on Build GRCh38
rs765714333 Chr.17: 47975836-47975836 on Build GRCh38
rs372191940 Chr.17: 47975839-47975839 on Build GRCh38
rs756247120 Chr.17: 47975933-47975933 on Build GRCh38
rs867486620 Chr.17: 47975975-47975975 on Build GRCh38
rs12948207 Chr.17: 47975987-47975987 on Build GRCh38
rs760961699 Chr.17: 47976001-47976001 on Build GRCh38
rs764542662 Chr.17: 47975842-47975842 on Build GRCh38
rs758964348 Chr.17: 47975981-47975981 on Build GRCh38
rs746001643 Chr.17: 47975865-47975865 on Build GRCh38
rs1008988189 Chr.17: 47975840-47975840 on Build GRCh38
rs201370626 Chr.17: 47976019-47976019 on Build GRCh38
rs200963209 Chr.17: 47975913-47975913 on Build GRCh38
rs764836302 Chr.17: 47975882-47975882 on Build GRCh38
rs950673912 Chr.17: 47975899-47975899 on Build GRCh38
rs756238130 Chr.17: 47975884-47975884 on Build GRCh38
rs771773882 Chr.17: 47975904-47975904 on Build GRCh38
rs368942321 Chr.17: 47975898-47975898 on Build GRCh38
rs202125432 Chr.17: 47975968-47975968 on Build GRCh38
rs915791047 Chr.17: 47975915-47975915 on Build GRCh38

View Full Product Details