Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45138966-45139219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD4
Entrez Gene ID
79777
Gene Name
acyl-CoA binding domain containing 4
Gene Aliases
HMFT0700
UniGene
Hs.110298
Gene Chromosome Location
Chr.17: 45132600-45144181 on Build GRCh38
Transcripts
NM_001135705.2 NM_001135706.2 NM_001135707.2 NM_001321352.1 NM_001321353.1 NM_024722.3 XM_006722085.2 XM_017025084.1 XM_017025085.1 XM_017025086.1 XM_017025087.1 XM_017025088.1 XM_017025089.1 XM_017025090.1 XM_017025091.1 XM_017025092.1 XM_017025093.1 XM_017025094.1 XM_017025095.1 XM_017025096.1 XM_017025097.1 XM_017025098.1 XM_017025099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596200 c.682G>A p.E228K Chr.17: 45139053-45139053 on Build GRCh38
4267858 c.767C>G p.A256G Chr.17: 45139100-45139100 on Build GRCh38
4130157 c.772A>G p.R258G Chr.17: 45139105-45139105 on Build GRCh38
1383778 c.769A>G p.M257V Chr.17: 45139140-45139140 on Build GRCh38
4267859 c.729C>G p.S243R Chr.17: 45139100-45139100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758358526 Chr.17: 45139121-45139121 on Build GRCh38
rs369014950 Chr.17: 45139042-45139042 on Build GRCh38
rs761016380 Chr.17: 45139101-45139101 on Build GRCh38
rs771241012 Chr.17: 45139164-45139164 on Build GRCh38
rs757604047 Chr.17: 45138993-45138997 on Build GRCh38
rs746706639 Chr.17: 45138989-45138989 on Build GRCh38
rs1052417977 Chr.17: 45139091-45139091 on Build GRCh38
rs200265169 Chr.17: 45139061-45139061 on Build GRCh38
rs745346160 Chr.17: 45139065-45139065 on Build GRCh38
rs557303418 Chr.17: 45139040-45139040 on Build GRCh38
rs377303243 Chr.17: 45139109-45139109 on Build GRCh38
rs776795835 Chr.17: 45139168-45139168 on Build GRCh38
rs759181684 Chr.17: 45139015-45139015 on Build GRCh38
rs780957691 Chr.17: 45139137-45139137 on Build GRCh38
rs770288651 Chr.17: 45139000-45139000 on Build GRCh38
rs374803518 Chr.17: 45139039-45139039 on Build GRCh38
rs777636266 Chr.17: 45139122-45139122 on Build GRCh38
rs772269471 Chr.17: 45139083-45139083 on Build GRCh38
rs568510716 Chr.17: 45139029-45139029 on Build GRCh38
rs769467191 Chr.17: 45139069-45139069 on Build GRCh38
rs758089830 Chr.17: 45139043-45139043 on Build GRCh38
rs753913162 Chr.17: 45139036-45139036 on Build GRCh38
rs751355553 Chr.17: 45139124-45139124 on Build GRCh38
rs748539089 Chr.17: 45139149-45139149 on Build GRCh38
rs766304951 Chr.17: 45139032-45139032 on Build GRCh38
rs774955396 Chr.17: 45139078-45139078 on Build GRCh38
rs540401788 Chr.17: 45139191-45139191 on Build GRCh38
rs773654105 Chr.17: 45139157-45139157 on Build GRCh38
rs759778665 Chr.17: 45139172-45139172 on Build GRCh38
rs772633053 Chr.17: 45139155-45139155 on Build GRCh38
rs201096011 Chr.17: 45139093-45139093 on Build GRCh38
rs761079435 Chr.17: 45139158-45139158 on Build GRCh38
rs756375848 Chr.17: 45139030-45139030 on Build GRCh38
rs1010966971 Chr.17: 45139103-45139103 on Build GRCh38
rs777636769 Chr.17: 45139054-45139054 on Build GRCh38
rs371382726 Chr.17: 45139151-45139151 on Build GRCh38
rs889253714 Chr.17: 45139108-45139108 on Build GRCh38
rs376543418 Chr.17: 45139062-45139062 on Build GRCh38
rs906453836 Chr.17: 45139073-45139073 on Build GRCh38
rs767951857 Chr.17: 45139020-45139020 on Build GRCh38
rs762235188 Chr.17: 45139017-45139017 on Build GRCh38
rs779720719 Chr.17: 45139148-45139148 on Build GRCh38
rs752768318 Chr.17: 45139188-45139188 on Build GRCh38
rs546790569 Chr.17: 45139016-45139016 on Build GRCh38
rs927216416 Chr.17: 45139174-45139174 on Build GRCh38
rs16939879 Chr.17: 45139105-45139105 on Build GRCh38
rs765233614 Chr.17: 45139175-45139175 on Build GRCh38
rs573177592 Chr.17: 45139146-45139146 on Build GRCh38
rs1009074243 Chr.17: 45139115-45139115 on Build GRCh38
rs757050209 Chr.17: 45139136-45139136 on Build GRCh38
rs776209925 Chr.17: 45139004-45139004 on Build GRCh38
rs769377488 Chr.17: 45139147-45139147 on Build GRCh38
rs752582690 Chr.17: 45139118-45139118 on Build GRCh38
rs762429455 Chr.17: 45139080-45139080 on Build GRCh38
rs765333936 Chr.17: 45139117-45139117 on Build GRCh38
rs369428697 Chr.17: 45139102-45139102 on Build GRCh38

View Full Product Details