Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99353870-99354131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC1A
Entrez Gene ID
10552
Gene Name
actin related protein 2/3 complex subunit 1A
Gene Aliases
Arc40, HEL-68, HEL-S-307, SOP2Hs, SOP2L
UniGene
Hs.124126
Gene Chromosome Location
Chr.7: 99325873-99366262 on Build GRCh38
Transcripts
NM_001190996.1 NM_006409.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4663476 c.653G>A p.R218H Chr.7: 99354061-99354061 on Build GRCh38
4788635 c.473G>T p.G158V Chr.7: 99354022-99354022 on Build GRCh38
4663477 c.512G>A p.R171H Chr.7: 99354061-99354061 on Build GRCh38
5835484 c.507G>C p.G169G Chr.7: 99354056-99354056 on Build GRCh38
1283510 c.666C>G p.V222V Chr.7: 99354074-99354074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145223474 Chr.7: 99354007-99354007 on Build GRCh38
rs372313254 Chr.7: 99354011-99354011 on Build GRCh38
rs376725540 Chr.7: 99354004-99354004 on Build GRCh38
rs146975353 Chr.7: 99354080-99354080 on Build GRCh38
rs11550144 Chr.7: 99354027-99354027 on Build GRCh38
rs752647954 Chr.7: 99353978-99353978 on Build GRCh38
rs773895160 Chr.7: 99354058-99354058 on Build GRCh38
rs73710441 Chr.7: 99354032-99354032 on Build GRCh38
rs781736363 Chr.7: 99354034-99354034 on Build GRCh38
rs770481583 Chr.7: 99354038-99354038 on Build GRCh38
rs764069129 Chr.7: 99354103-99354103 on Build GRCh38
rs775050152 Chr.7: 99354089-99354089 on Build GRCh38
rs767022179 Chr.7: 99354081-99354081 on Build GRCh38
rs952422659 Chr.7: 99353949-99353949 on Build GRCh38
rs773688364 Chr.7: 99354051-99354051 on Build GRCh38
rs558106225 Chr.7: 99353960-99353960 on Build GRCh38
rs894318200 Chr.7: 99354092-99354092 on Build GRCh38
rs756403062 Chr.7: 99353929-99353929 on Build GRCh38
rs539217463 Chr.7: 99353957-99353957 on Build GRCh38
rs765993633 Chr.7: 99353946-99353946 on Build GRCh38
rs748658169 Chr.7: 99354036-99354036 on Build GRCh38
rs571833149 Chr.7: 99353925-99353925 on Build GRCh38
rs758625084 Chr.7: 99354008-99354008 on Build GRCh38
rs150009166 Chr.7: 99353989-99353989 on Build GRCh38
rs760596520 Chr.7: 99354090-99354090 on Build GRCh38
rs573083020 Chr.7: 99353976-99353976 on Build GRCh38
rs768642928 Chr.7: 99354033-99354033 on Build GRCh38
rs763068686 Chr.7: 99353897-99353897 on Build GRCh38
rs914625005 Chr.7: 99354017-99354017 on Build GRCh38
rs200602020 Chr.7: 99354104-99354104 on Build GRCh38
rs147646339 Chr.7: 99354097-99354097 on Build GRCh38
rs763263013 Chr.7: 99354053-99354053 on Build GRCh38
rs770573211 Chr.7: 99354056-99354056 on Build GRCh38

View Full Product Details