Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 37472687-37472918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK1A
Entrez Gene ID
1859
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1A
Gene Aliases
DYRK, DYRK1, HP86, MNB, MNBH, MRD7
UniGene
Hs.368240
Gene Chromosome Location
Chr.21: 37365790-37515376 on Build GRCh38
Transcripts
NM_001396.3 NM_101395.2 NM_130436.2 NM_130438.2 XM_006723976.3 XM_006723977.3 XM_006723978.3 XM_006723979.3 XM_011529482.2 XM_011529483.2 XM_011529484.2 XM_011529485.2 XM_017028284.1 XM_017028285.1 XM_017028286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6028227 c.170_176delCATATTC p.S57fs*8 Chr.21: 37472843-37472849 on Build GRCh38
5217729 c.46C>T p.R16W Chr.21: 37472719-37472719 on Build GRCh38
6250986 c.131G>T p.R44L Chr.21: 37472804-37472804 on Build GRCh38
5887522 c.82C>T p.L28F Chr.21: 37472755-37472755 on Build GRCh38
5834028 c.183G>C p.Q61H Chr.21: 37472856-37472856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968549141 Chr.21: 37472776-37472776 on Build GRCh38
rs1041691170 Chr.21: 37472846-37472846 on Build GRCh38
rs776043893 Chr.21: 37472766-37472766 on Build GRCh38
rs758573762 Chr.21: 37472803-37472803 on Build GRCh38
rs896508492 Chr.21: 37472751-37472751 on Build GRCh38
rs1010198969 Chr.21: 37472760-37472760 on Build GRCh38
rs367984873 Chr.21: 37472800-37472800 on Build GRCh38
rs771919737 Chr.21: 37472713-37472713 on Build GRCh38
rs745895020 Chr.21: 37472712-37472712 on Build GRCh38
rs1057175147 Chr.21: 37472720-37472720 on Build GRCh38
rs1049761 Chr.21: 37472826-37472826 on Build GRCh38
rs750600368 Chr.21: 37472801-37472801 on Build GRCh38
rs764466157 Chr.21: 37472778-37472778 on Build GRCh38
rs754161665 Chr.21: 37472795-37472795 on Build GRCh38
rs765791729 Chr.21: 37472764-37472764 on Build GRCh38
rs1056885794 Chr.21: 37472729-37472729 on Build GRCh38
rs762466544 Chr.21: 37472761-37472761 on Build GRCh38
rs368821516 Chr.21: 37472871-37472871 on Build GRCh38
rs1049754 Chr.21: 37472733-37472733 on Build GRCh38
rs1049755 Chr.21: 37472736-37472736 on Build GRCh38
rs747907599 Chr.21: 37472844-37472844 on Build GRCh38
rs536947921 Chr.21: 37472748-37472748 on Build GRCh38
rs1049757 Chr.21: 37472781-37472781 on Build GRCh38
rs773802285 Chr.21: 37472877-37472877 on Build GRCh38
rs1049760 Chr.21: 37472813-37472813 on Build GRCh38
rs780254690 Chr.21: 37472804-37472804 on Build GRCh38
rs772599209 Chr.21: 37472758-37472758 on Build GRCh38
rs1049758 Chr.21: 37472787-37472787 on Build GRCh38
rs1049759 Chr.21: 37472811-37472811 on Build GRCh38
rs769342483 Chr.21: 37472850-37472850 on Build GRCh38
rs587776929 Chr.21: 37472815-37472816 on Build GRCh38
rs1018001317 Chr.21: 37472865-37472865 on Build GRCh38
rs747774015 Chr.21: 37472730-37472730 on Build GRCh38
rs776541736 Chr.21: 37472728-37472728 on Build GRCh38
rs768615289 Chr.21: 37472723-37472723 on Build GRCh38
rs903016731 Chr.21: 37472870-37472870 on Build GRCh38
rs1049763 Chr.21: 37472842-37472842 on Build GRCh38
rs1049756 Chr.21: 37472768-37472768 on Build GRCh38
rs201180170 Chr.21: 37472769-37472769 on Build GRCh38
rs754909260 Chr.21: 37472841-37472841 on Build GRCh38
rs780200883 Chr.21: 37472716-37472716 on Build GRCh38
rs748888299 Chr.21: 37472862-37472862 on Build GRCh38
rs1049753 Chr.21: 37472718-37472718 on Build GRCh38
rs769445831 Chr.21: 37472752-37472752 on Build GRCh38
rs373178770 Chr.21: 37472860-37472860 on Build GRCh38
rs746988114 Chr.21: 37472835-37472835 on Build GRCh38
rs1049762 Chr.21: 37472832-37472832 on Build GRCh38

View Full Product Details