Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113729204-113729473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP7
Entrez Gene ID
840
Gene Name
caspase 7
Gene Aliases
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3
UniGene
Hs.9216
Gene Chromosome Location
Chr.10: 113679162-113730909 on Build GRCh38
Transcripts
NM_001227.4 NM_001267056.1 NM_001267057.1 NM_001267058.1 NM_001320911.1 NM_033338.5 NM_033339.4 NM_033340.3 XM_006718017.3 XM_011540260.1 XM_017016763.1 XM_017016764.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3434232 c.802C>T p.P268S Chr.10: 113729331-113729331 on Build GRCh38
7216839 c.864C>G p.D288E Chr.10: 113729393-113729393 on Build GRCh38
3434231 c.791C>T p.S264L Chr.10: 113729320-113729320 on Build GRCh38
7215507 c.879G>A p.Q293Q Chr.10: 113729408-113729408 on Build GRCh38
1134966 c.796A>T p.R266W Chr.10: 113729325-113729325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776647432 Chr.10: 113729451-113729451 on Build GRCh38
rs745988255 Chr.10: 113729303-113729303 on Build GRCh38
rs781723563 Chr.10: 113729417-113729417 on Build GRCh38
rs61755278 Chr.10: 113729428-113729428 on Build GRCh38
rs868234989 Chr.10: 113729365-113729365 on Build GRCh38
rs967141128 Chr.10: 113729245-113729245 on Build GRCh38
rs773169485 Chr.10: 113729357-113729357 on Build GRCh38
rs2227309 Chr.10: 113729408-113729408 on Build GRCh38
rs141834215 Chr.10: 113729298-113729298 on Build GRCh38
rs974230032 Chr.10: 113729280-113729280 on Build GRCh38
rs2227310 Chr.10: 113729393-113729393 on Build GRCh38
rs1038826941 Chr.10: 113729386-113729386 on Build GRCh38
rs1127684 Chr.10: 113729321-113729321 on Build GRCh38
rs757003092 Chr.10: 113729291-113729295 on Build GRCh38
rs778568829 Chr.10: 113729276-113729276 on Build GRCh38
rs116437863 Chr.10: 113729418-113729418 on Build GRCh38
rs997856753 Chr.10: 113729397-113729397 on Build GRCh38
rs749560623 Chr.10: 113729436-113729436 on Build GRCh38
rs908586546 Chr.10: 113729234-113729234 on Build GRCh38
rs748448530 Chr.10: 113729285-113729285 on Build GRCh38
rs903604697 Chr.10: 113729356-113729356 on Build GRCh38
rs1042128768 Chr.10: 113729355-113729355 on Build GRCh38
rs112264603 Chr.10: 113729339-113729339 on Build GRCh38
rs545011125 Chr.10: 113729375-113729375 on Build GRCh38
rs771971818 Chr.10: 113729438-113729438 on Build GRCh38
rs770709414 Chr.10: 113729449-113729449 on Build GRCh38
rs531534527 Chr.10: 113729446-113729446 on Build GRCh38
rs778154041 Chr.10: 113729396-113729396 on Build GRCh38
rs780420016 Chr.10: 113729432-113729432 on Build GRCh38
rs770071040 Chr.10: 113729314-113729314 on Build GRCh38
rs534130735 Chr.10: 113729259-113729259 on Build GRCh38
rs775867698 Chr.10: 113729320-113729320 on Build GRCh38
rs147969141 Chr.10: 113729288-113729288 on Build GRCh38
rs914055312 Chr.10: 113729228-113729228 on Build GRCh38
rs146349235 Chr.10: 113729258-113729258 on Build GRCh38
rs191429899 Chr.10: 113729369-113729369 on Build GRCh38
rs754523047 Chr.10: 113729384-113729384 on Build GRCh38
rs772834414 Chr.10: 113729443-113729443 on Build GRCh38
rs752406752 Chr.10: 113729390-113729390 on Build GRCh38
rs776522239 Chr.10: 113729300-113729300 on Build GRCh38
rs974261027 Chr.10: 113729444-113729444 on Build GRCh38
rs764815610 Chr.10: 113729385-113729385 on Build GRCh38
rs200326742 Chr.10: 113729328-113729328 on Build GRCh38
rs758779463 Chr.10: 113729287-113729287 on Build GRCh38
rs760579502 Chr.10: 113729368-113729368 on Build GRCh38
rs1032292028 Chr.10: 113729450-113729450 on Build GRCh38
rs747418413 Chr.10: 113729403-113729403 on Build GRCh38
rs747539394 Chr.10: 113729291-113729291 on Build GRCh38

View Full Product Details