Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127599324-127599597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCN3
Entrez Gene ID
29999
Gene Name
fascin actin-bundling protein 3
Gene Aliases
-
UniGene
Hs.712692
Gene Chromosome Location
Chr.7: 127593635-127601797 on Build GRCh38
Transcripts
NM_020369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6483942 c.1196G>T p.G399V Chr.7: 127599456-127599456 on Build GRCh38
2768616 c.1180C>T p.R394C Chr.7: 127599440-127599440 on Build GRCh38
7236962 c.1145T>A p.L382* Chr.7: 127599405-127599405 on Build GRCh38
4568659 c.1264T>A p.C422S Chr.7: 127599524-127599524 on Build GRCh38
311308 c.1157G>T p.R386L Chr.7: 127599417-127599417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754833506 Chr.7: 127599574-127599574 on Build GRCh38
rs763779206 Chr.7: 127599483-127599483 on Build GRCh38
rs758047596 Chr.7: 127599481-127599481 on Build GRCh38
rs762051152 Chr.7: 127599450-127599450 on Build GRCh38
rs773868040 Chr.7: 127599462-127599462 on Build GRCh38
rs199739794 Chr.7: 127599525-127599525 on Build GRCh38
rs148039274 Chr.7: 127599502-127599502 on Build GRCh38
rs774540624 Chr.7: 127599445-127599445 on Build GRCh38
rs762870630 Chr.7: 127599376-127599376 on Build GRCh38
rs753652855 Chr.7: 127599571-127599571 on Build GRCh38
rs752242330 Chr.7: 127599479-127599479 on Build GRCh38
rs769905535 Chr.7: 127599347-127599347 on Build GRCh38
rs763725735 Chr.7: 127599532-127599532 on Build GRCh38
rs141748303 Chr.7: 127599503-127599503 on Build GRCh38
rs536433642 Chr.7: 127599491-127599491 on Build GRCh38
rs201189518 Chr.7: 127599414-127599414 on Build GRCh38
rs147838592 Chr.7: 127599397-127599397 on Build GRCh38
rs140420927 Chr.7: 127599500-127599500 on Build GRCh38
rs370690397 Chr.7: 127599440-127599440 on Build GRCh38
rs768122917 Chr.7: 127599456-127599456 on Build GRCh38
rs748179643 Chr.7: 127599514-127599514 on Build GRCh38
rs146973835 Chr.7: 127599466-127599466 on Build GRCh38
rs768804902 Chr.7: 127599444-127599444 on Build GRCh38
rs781053262 Chr.7: 127599496-127599496 on Build GRCh38
rs377654536 Chr.7: 127599419-127599419 on Build GRCh38
rs774771663 Chr.7: 127599507-127599507 on Build GRCh38
rs34941808 Chr.7: 127599531-127599531 on Build GRCh38
rs938053495 Chr.7: 127599430-127599430 on Build GRCh38
rs201240741 Chr.7: 127599528-127599528 on Build GRCh38
rs184679485 Chr.7: 127599527-127599527 on Build GRCh38
rs531153218 Chr.7: 127599387-127599387 on Build GRCh38
rs773553881 Chr.7: 127599522-127599522 on Build GRCh38
rs534616686 Chr.7: 127599554-127599554 on Build GRCh38
rs375094249 Chr.7: 127599350-127599350 on Build GRCh38
rs369301871 Chr.7: 127599359-127599359 on Build GRCh38
rs777288563 Chr.7: 127599420-127599420 on Build GRCh38
rs202131198 Chr.7: 127599435-127599435 on Build GRCh38
rs147048987 Chr.7: 127599380-127599380 on Build GRCh38
rs373334080 Chr.7: 127599441-127599441 on Build GRCh38
rs188307221 Chr.7: 127599429-127599429 on Build GRCh38
rs761250404 Chr.7: 127599541-127599541 on Build GRCh38
rs761269365 Chr.7: 127599524-127599524 on Build GRCh38
rs746572057 Chr.7: 127599426-127599426 on Build GRCh38
rs927919464 Chr.7: 127599526-127599526 on Build GRCh38
rs150488515 Chr.7: 127599506-127599506 on Build GRCh38
rs772282312 Chr.7: 127599517-127599517 on Build GRCh38
rs569040345 Chr.7: 127599570-127599570 on Build GRCh38
rs768178179 Chr.7: 127599405-127599405 on Build GRCh38
rs369121584 Chr.7: 127599416-127599416 on Build GRCh38
rs752400150 Chr.7: 127599417-127599417 on Build GRCh38
rs34394613 Chr.7: 127599543-127599543 on Build GRCh38
rs1046505335 Chr.7: 127599406-127599406 on Build GRCh38
rs751230329 Chr.7: 127599534-127599534 on Build GRCh38
rs35833269 Chr.7: 127599465-127599465 on Build GRCh38
rs192546949 Chr.7: 127599434-127599434 on Build GRCh38
rs756044191 Chr.7: 127599563-127599563 on Build GRCh38

View Full Product Details