Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121621711-121621962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO40
Entrez Gene ID
51725
Gene Name
F-box protein 40
Gene Aliases
FBX40
UniGene
Hs.272564
Gene Chromosome Location
Chr.3: 121593323-121630292 on Build GRCh38
Transcripts
NM_016298.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
479283 c.496G>T p.V166L Chr.3: 121621925-121621925 on Build GRCh38
4760730 c.485G>A p.G162D Chr.3: 121621914-121621914 on Build GRCh38
5626275 c.315G>A p.V105V Chr.3: 121621744-121621744 on Build GRCh38
4842176 c.487G>A p.E163K Chr.3: 121621916-121621916 on Build GRCh38
4827466 c.508G>A p.G170R Chr.3: 121621937-121621937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377409069 Chr.3: 121621786-121621786 on Build GRCh38
rs903106868 Chr.3: 121621748-121621748 on Build GRCh38
rs372552970 Chr.3: 121621931-121621931 on Build GRCh38
rs909232803 Chr.3: 121621878-121621878 on Build GRCh38
rs946645796 Chr.3: 121621758-121621758 on Build GRCh38
rs748062583 Chr.3: 121621816-121621816 on Build GRCh38
rs200154550 Chr.3: 121621901-121621901 on Build GRCh38
rs748921319 Chr.3: 121621880-121621880 on Build GRCh38
rs904817973 Chr.3: 121621771-121621771 on Build GRCh38
rs775065669 Chr.3: 121621784-121621784 on Build GRCh38
rs777845129 Chr.3: 121621743-121621743 on Build GRCh38
rs757299930 Chr.3: 121621858-121621858 on Build GRCh38
rs1007346895 Chr.3: 121621791-121621791 on Build GRCh38
rs760595092 Chr.3: 121621897-121621897 on Build GRCh38
rs774121135 Chr.3: 121621883-121621883 on Build GRCh38
rs938025587 Chr.3: 121621888-121621888 on Build GRCh38
rs770193085 Chr.3: 121621783-121621783 on Build GRCh38
rs752867730 Chr.3: 121621817-121621817 on Build GRCh38
rs745710712 Chr.3: 121621734-121621734 on Build GRCh38
rs997067161 Chr.3: 121621919-121621919 on Build GRCh38
rs770070135 Chr.3: 121621868-121621868 on Build GRCh38
rs767611473 Chr.3: 121621799-121621799 on Build GRCh38
rs549329473 Chr.3: 121621835-121621835 on Build GRCh38
rs761861151 Chr.3: 121621798-121621798 on Build GRCh38
rs976661978 Chr.3: 121621832-121621832 on Build GRCh38
rs746363538 Chr.3: 121621865-121621865 on Build GRCh38
rs775854273 Chr.3: 121621873-121621873 on Build GRCh38
rs915181976 Chr.3: 121621735-121621735 on Build GRCh38
rs759230547 Chr.3: 121621921-121621921 on Build GRCh38
rs757387582 Chr.3: 121621940-121621940 on Build GRCh38
rs766181328 Chr.3: 121621815-121621815 on Build GRCh38
rs1017101413 Chr.3: 121621814-121621814 on Build GRCh38
rs753621120 Chr.3: 121621920-121621920 on Build GRCh38
rs747546878 Chr.3: 121621759-121621759 on Build GRCh38
rs756038658 Chr.3: 121621809-121621809 on Build GRCh38
rs771379652 Chr.3: 121621764-121621764 on Build GRCh38
rs748396801 Chr.3: 121621730-121621730 on Build GRCh38
rs763515142 Chr.3: 121621787-121621787 on Build GRCh38
rs267599566 Chr.3: 121621848-121621848 on Build GRCh38
rs1056199005 Chr.3: 121621788-121621788 on Build GRCh38
rs772318817 Chr.3: 121621762-121621762 on Build GRCh38
rs566081092 Chr.3: 121621885-121621885 on Build GRCh38
rs777104772 Chr.3: 121621770-121621770 on Build GRCh38
rs375428007 Chr.3: 121621881-121621881 on Build GRCh38
rs767131081 Chr.3: 121621889-121621889 on Build GRCh38
rs781193078 Chr.3: 121621941-121621941 on Build GRCh38
rs758423510 Chr.3: 121621731-121621731 on Build GRCh38
rs370258531 Chr.3: 121621806-121621806 on Build GRCh38
rs752659519 Chr.3: 121621812-121621812 on Build GRCh38
rs34751480 Chr.3: 121621796-121621796 on Build GRCh38
rs142028632 Chr.3: 121621773-121621773 on Build GRCh38
rs570315447 Chr.3: 121621745-121621745 on Build GRCh38
rs199575816 Chr.3: 121621928-121621928 on Build GRCh38
rs138735736 Chr.3: 121621849-121621849 on Build GRCh38
rs773338926 Chr.3: 121621894-121621894 on Build GRCh38

View Full Product Details