Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 56193293-56193518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85A
Entrez Gene ID
114800
Gene Name
coiled-coil domain containing 85A
Gene Aliases
-
UniGene
Hs.117136
Gene Chromosome Location
Chr.2: 56184123-56386174 on Build GRCh38
Transcripts
NM_001080433.1 XM_005264125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5619567 c.1166G>T p.R389M Chr.2: 56193366-56193366 on Build GRCh38
721461 c.1123G>A p.G375R Chr.2: 56193323-56193323 on Build GRCh38
3991379 c.1156G>A p.G386R Chr.2: 56193356-56193356 on Build GRCh38
4459216 c.1116C>T p.H372H Chr.2: 56193316-56193316 on Build GRCh38
721460 c.1195G>A p.E399K Chr.2: 56193395-56193395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767051265 Chr.2: 56193448-56193448 on Build GRCh38
rs199630451 Chr.2: 56193337-56193337 on Build GRCh38
rs762184153 Chr.2: 56193332-56193340 on Build GRCh38
rs368722854 Chr.2: 56193422-56193422 on Build GRCh38
rs767436502 Chr.2: 56193342-56193342 on Build GRCh38
rs114846283 Chr.2: 56193416-56193416 on Build GRCh38
rs551475492 Chr.2: 56193348-56193348 on Build GRCh38
rs372992628 Chr.2: 56193323-56193323 on Build GRCh38
rs764667803 Chr.2: 56193492-56193492 on Build GRCh38
rs548105160 Chr.2: 56193470-56193470 on Build GRCh38
rs777550693 Chr.2: 56193379-56193379 on Build GRCh38
rs746576522 Chr.2: 56193381-56193381 on Build GRCh38
rs373804374 Chr.2: 56193484-56193484 on Build GRCh38
rs767663265 Chr.2: 56193427-56193427 on Build GRCh38
rs780793877 Chr.2: 56193481-56193481 on Build GRCh38
rs370347176 Chr.2: 56193483-56193483 on Build GRCh38
rs773463689 Chr.2: 56193434-56193434 on Build GRCh38
rs775228056 Chr.2: 56193318-56193318 on Build GRCh38
rs745840613 Chr.2: 56193398-56193398 on Build GRCh38
rs756647145 Chr.2: 56193352-56193352 on Build GRCh38
rs139777024 Chr.2: 56193349-56193349 on Build GRCh38
rs756823201 Chr.2: 56193480-56193480 on Build GRCh38
rs1045327577 Chr.2: 56193358-56193358 on Build GRCh38
rs895053148 Chr.2: 56193393-56193393 on Build GRCh38
rs759792708 Chr.2: 56193370-56193370 on Build GRCh38
rs775387962 Chr.2: 56193403-56193403 on Build GRCh38
rs374067476 Chr.2: 56193383-56193383 on Build GRCh38
rs1057253111 Chr.2: 56193409-56193409 on Build GRCh38
rs533959630 Chr.2: 56193333-56193333 on Build GRCh38
rs926744834 Chr.2: 56193360-56193360 on Build GRCh38
rs760813992 Chr.2: 56193439-56193439 on Build GRCh38
rs754303873 Chr.2: 56193362-56193362 on Build GRCh38
rs749168116 Chr.2: 56193411-56193411 on Build GRCh38
rs768817902 Chr.2: 56193461-56193461 on Build GRCh38
rs754391823 Chr.2: 56193459-56193459 on Build GRCh38
rs780632509 Chr.2: 56193390-56193390 on Build GRCh38
rs769765007 Chr.2: 56193401-56193401 on Build GRCh38
rs144495274 Chr.2: 56193491-56193491 on Build GRCh38
rs762709570 Chr.2: 56193322-56193322 on Build GRCh38
rs766959547 Chr.2: 56193356-56193356 on Build GRCh38
rs368840161 Chr.2: 56193485-56193485 on Build GRCh38

View Full Product Details