Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118349874-118350103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3G
Entrez Gene ID
917
Gene Name
CD3g molecule
Gene Aliases
CD3-GAMMA, IMD17, T3G
UniGene
Hs.2259
Gene Chromosome Location
Chr.11: 118344307-118353782 on Build GRCh38
Transcripts
NM_000073.2 XM_005271724.3 XM_006718941.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923542 c.223G>A p.G75R Chr.11: 118349886-118349886 on Build GRCh38
5522344 c.216G>A p.W72* Chr.11: 118349879-118349879 on Build GRCh38
3443920 c.266G>A p.G89E Chr.11: 118349929-118349929 on Build GRCh38
923543 c.305G>C p.R102T Chr.11: 118349968-118349968 on Build GRCh38
5831665 c.306delA p.M103fs*8 Chr.11: 118349969-118349969 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1042064987 Chr.11: 118349904-118349904 on Build GRCh38
rs201366722 Chr.11: 118349968-118349968 on Build GRCh38
rs370770359 Chr.11: 118349975-118349975 on Build GRCh38
rs201795915 Chr.11: 118349957-118349957 on Build GRCh38
rs201124887 Chr.11: 118349907-118349907 on Build GRCh38
rs200028255 Chr.11: 118349906-118349906 on Build GRCh38
rs566420322 Chr.11: 118350058-118350058 on Build GRCh38
rs202047276 Chr.11: 118349992-118349992 on Build GRCh38
rs201524785 Chr.11: 118349921-118349921 on Build GRCh38
rs760311198 Chr.11: 118350001-118350001 on Build GRCh38
rs748368894 Chr.11: 118349977-118349977 on Build GRCh38
rs758764358 Chr.11: 118349908-118349908 on Build GRCh38
rs776875720 Chr.11: 118349959-118349959 on Build GRCh38
rs780136472 Chr.11: 118349916-118349916 on Build GRCh38
rs200125722 Chr.11: 118349923-118349923 on Build GRCh38
rs767318320 Chr.11: 118349991-118349991 on Build GRCh38
rs200487871 Chr.11: 118349958-118349958 on Build GRCh38
rs770822390 Chr.11: 118349980-118349980 on Build GRCh38
rs201964905 Chr.11: 118349983-118349983 on Build GRCh38
rs141052321 Chr.11: 118349936-118349936 on Build GRCh38
rs759524595 Chr.11: 118349987-118349987 on Build GRCh38
rs1014014116 Chr.11: 118349999-118349999 on Build GRCh38
rs1033882270 Chr.11: 118349938-118349938 on Build GRCh38
rs186234121 Chr.11: 118350050-118350050 on Build GRCh38
rs693187 Chr.11: 118350067-118350067 on Build GRCh38
rs76479097 Chr.11: 118350061-118350061 on Build GRCh38
rs1005814372 Chr.11: 118349970-118349970 on Build GRCh38
rs758659595 Chr.11: 118349905-118349905 on Build GRCh38

View Full Product Details