Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113652726-113652957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4B
Entrez Gene ID
496
Gene Name
ATPase H+/K+ transporting beta subunit
Gene Aliases
ATP6B
UniGene
Hs.434202
Gene Chromosome Location
Chr.13: 113648804-113658198 on Build GRCh38
Transcripts
NM_000705.3 XM_017020684.1

Gene Symbol
GRK1
Entrez Gene ID
6011
Gene Name
G protein-coupled receptor kinase 1
Gene Aliases
GPRK1, RHOK, RK
UniGene
Hs.103501
Gene Chromosome Location
Chr.13: 113645771-113735664 on Build GRCh38
Transcripts
NM_000705.3 XM_017020684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589762 c.490G>A p.G164S Chr.13: 113652938-113652938 on Build GRCh38
3955422 c.493C>A p.L165M Chr.13: 113652935-113652935 on Build GRCh38
80896 c.504C>G p.P168P Chr.13: 113652924-113652924 on Build GRCh38
2264464 c.524G>A p.G175E Chr.13: 113652904-113652904 on Build GRCh38
695921 c.520G>T p.E174* Chr.13: 113652908-113652908 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375234995 Chr.13: 113652879-113652879 on Build GRCh38
rs758073640 Chr.13: 113652923-113652923 on Build GRCh38
rs778344246 Chr.13: 113652838-113652839 on Build GRCh38
rs1027239357 Chr.13: 113652763-113652763 on Build GRCh38
rs750101696 Chr.13: 113652883-113652884 on Build GRCh38
rs780631070 Chr.13: 113652756-113652756 on Build GRCh38
rs770890534 Chr.13: 113652776-113652776 on Build GRCh38
rs756791181 Chr.13: 113652823-113652824 on Build GRCh38
rs762089260 Chr.13: 113652856-113652856 on Build GRCh38
rs764093280 Chr.13: 113652801-113652801 on Build GRCh38
rs778919864 Chr.13: 113652911-113652911 on Build GRCh38
rs571742002 Chr.13: 113652813-113652813 on Build GRCh38
rs780066459 Chr.13: 113652930-113652930 on Build GRCh38
rs1056828188 Chr.13: 113652929-113652929 on Build GRCh38
rs776873874 Chr.13: 113652830-113652830 on Build GRCh38
rs1044194818 Chr.13: 113652877-113652877 on Build GRCh38
rs750837224 Chr.13: 113652918-113652918 on Build GRCh38
rs370368571 Chr.13: 113652872-113652872 on Build GRCh38
rs776675253 Chr.13: 113652933-113652933 on Build GRCh38
rs745851797 Chr.13: 113652778-113652778 on Build GRCh38
rs753396275 Chr.13: 113652791-113652792 on Build GRCh38
rs1017631562 Chr.13: 113652788-113652788 on Build GRCh38
rs765644944 Chr.13: 113652844-113652844 on Build GRCh38
rs757920207 Chr.13: 113652897-113652898 on Build GRCh38
rs775226881 Chr.13: 113652809-113652809 on Build GRCh38
rs753594200 Chr.13: 113652875-113652875 on Build GRCh38
rs772044761 Chr.13: 113652797-113652797 on Build GRCh38
rs185676558 Chr.13: 113652885-113652885 on Build GRCh38
rs886265695 Chr.13: 113652786-113652786 on Build GRCh38
rs745682358 Chr.13: 113652917-113652917 on Build GRCh38
rs763666844 Chr.13: 113652874-113652874 on Build GRCh38
rs913014127 Chr.13: 113652869-113652869 on Build GRCh38
rs377373086 Chr.13: 113652857-113652857 on Build GRCh38
rs976073179 Chr.13: 113652803-113652803 on Build GRCh38
rs768677010 Chr.13: 113652829-113652829 on Build GRCh38
rs116830694 Chr.13: 113652847-113652847 on Build GRCh38
rs373566809 Chr.13: 113652846-113652846 on Build GRCh38
rs994066935 Chr.13: 113652751-113652751 on Build GRCh38
rs767713017 Chr.13: 113652865-113652865 on Build GRCh38
rs761996576 Chr.13: 113652839-113652839 on Build GRCh38
rs748432358 Chr.13: 113652935-113652935 on Build GRCh38
rs532281305 Chr.13: 113652758-113652758 on Build GRCh38
rs193141905 Chr.13: 113652764-113652764 on Build GRCh38
rs992584457 Chr.13: 113652860-113652860 on Build GRCh38
rs75160036 Chr.13: 113652931-113652931 on Build GRCh38
rs138473007 Chr.13: 113652765-113652765 on Build GRCh38
rs963291077 Chr.13: 113652819-113652819 on Build GRCh38

View Full Product Details