Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48894229-48894488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5G1
Entrez Gene ID
516
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9)
Gene Aliases
ATP5A, ATP5G
UniGene
Hs.80986
Gene Chromosome Location
Chr.17: 48892786-48895871 on Build GRCh38
Transcripts
NM_001002027.1 NM_005175.2 NR_135674.1

Gene Symbol
LOC105371814
Entrez Gene ID
105371814
Gene Name
uncharacterized LOC105371814
Gene Aliases
-
UniGene
Hs.655254
Gene Chromosome Location
Chr.17: 48874860-48908983 on Build GRCh38
Transcripts
NM_001002027.1 NM_005175.2 NR_135674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4623596 c.45C>G p.R15R Chr.17: 48894377-48894377 on Build GRCh38
309368 c.40-2A>G p.? Chr.17: 48894370-48894370 on Build GRCh38
6435096 c.89T>C p.L30S Chr.17: 48894421-48894421 on Build GRCh38
2697950 c.56G>A p.R19K Chr.17: 48894388-48894388 on Build GRCh38
405032 c.42delC p.R15fs*7 Chr.17: 48894374-48894374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201426120 Chr.17: 48894375-48894375 on Build GRCh38
rs534058266 Chr.17: 48894323-48894323 on Build GRCh38
rs758016938 Chr.17: 48894376-48894376 on Build GRCh38
rs755312179 Chr.17: 48894434-48894434 on Build GRCh38
rs1803174 Chr.17: 48894418-48894418 on Build GRCh38
rs371950583 Chr.17: 48894361-48894361 on Build GRCh38
rs974564377 Chr.17: 48894332-48894332 on Build GRCh38
rs11549738 Chr.17: 48894399-48894399 on Build GRCh38
rs749727998 Chr.17: 48894416-48894416 on Build GRCh38
rs559625015 Chr.17: 48894336-48894336 on Build GRCh38
rs763726177 Chr.17: 48894383-48894383 on Build GRCh38
rs964910984 Chr.17: 48894328-48894328 on Build GRCh38
rs113959540 Chr.17: 48894342-48894342 on Build GRCh38
rs779288492 Chr.17: 48894444-48894444 on Build GRCh38
rs780692172 Chr.17: 48894398-48894398 on Build GRCh38
rs748350954 Chr.17: 48894458-48894458 on Build GRCh38
rs966582350 Chr.17: 48894403-48894403 on Build GRCh38
rs11549735 Chr.17: 48894435-48894435 on Build GRCh38
rs751080645 Chr.17: 48894388-48894388 on Build GRCh38
rs907545065 Chr.17: 48894373-48894373 on Build GRCh38
rs112530546 Chr.17: 48894370-48894370 on Build GRCh38
rs766284135 Chr.17: 48894353-48894353 on Build GRCh38
rs752301557 Chr.17: 48894367-48894367 on Build GRCh38
rs777869807 Chr.17: 48894388-48894388 on Build GRCh38
rs756314540 Chr.17: 48894322-48894323 on Build GRCh38
rs369828692 Chr.17: 48894356-48894356 on Build GRCh38
rs756732763 Chr.17: 48894389-48894389 on Build GRCh38
rs772252604 Chr.17: 48894464-48894464 on Build GRCh38
rs767273094 Chr.17: 48894297-48894297 on Build GRCh38
rs983102437 Chr.17: 48894372-48894372 on Build GRCh38
rs528434194 Chr.17: 48894345-48894345 on Build GRCh38

View Full Product Details