Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 95101526-95101799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
UniGene
Hs.434253
Gene Chromosome Location
Chr.9: 94726604-95148264 on Build GRCh38
Transcripts
XM_011519121.2 NM_000136.2 NM_001243743.1 XM_005251802.3 XM_006717001.2 XM_011518365.2 XM_011518367.2 XM_017014452.1 XM_017014453.1 XM_017014454.1

Gene Symbol
FANCC
Entrez Gene ID
2176
Gene Name
Fanconi anemia complementation group C
Gene Aliases
FA3, FAC, FACC
UniGene
Hs.494529
Gene Chromosome Location
Chr.9: 95099054-95317730 on Build GRCh38
Transcripts
XM_011519121.2 NM_000136.2 NM_001243743.1 XM_005251802.3 XM_006717001.2 XM_011518365.2 XM_011518367.2 XM_017014452.1 XM_017014453.1 XM_017014454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6958651 c.1619G>A p.S540N Chr.9: 95101765-95101765 on Build GRCh38
6255835 c.1628C>A p.S543* Chr.9: 95101756-95101756 on Build GRCh38
1625273 c.1617A>C p.E539D Chr.9: 95101767-95101767 on Build GRCh38
138719 c.1599G>A p.W533* Chr.9: 95101785-95101785 on Build GRCh38
396352 c.1674C>A p.V558V Chr.9: 95101710-95101710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189154697 Chr.9: 95101564-95101564 on Build GRCh38
rs750866823 Chr.9: 95101709-95101709 on Build GRCh38
rs867319477 Chr.9: 95101756-95101756 on Build GRCh38
rs976861230 Chr.9: 95101615-95101615 on Build GRCh38
rs104886457 Chr.9: 95101742-95101742 on Build GRCh38
rs1026925048 Chr.9: 95101590-95101590 on Build GRCh38
rs7048910 Chr.9: 95101591-95101591 on Build GRCh38
rs529828813 Chr.9: 95101625-95101625 on Build GRCh38
rs375908568 Chr.9: 95101677-95101677 on Build GRCh38
rs34760720 Chr.9: 95101669-95101669 on Build GRCh38
rs531605962 Chr.9: 95101678-95101678 on Build GRCh38
rs926261922 Chr.9: 95101634-95101634 on Build GRCh38
rs1015228314 Chr.9: 95101697-95101697 on Build GRCh38
rs1027358273 Chr.9: 95101773-95101773 on Build GRCh38
rs757786105 Chr.9: 95101660-95101660 on Build GRCh38
rs771289098 Chr.9: 95101596-95101596 on Build GRCh38
rs775336408 Chr.9: 95101692-95101692 on Build GRCh38
rs747229495 Chr.9: 95101671-95101671 on Build GRCh38
rs864622417 Chr.9: 95101756-95101757 on Build GRCh38
rs117175949 Chr.9: 95101702-95101702 on Build GRCh38
rs774109877 Chr.9: 95101683-95101683 on Build GRCh38
rs964425118 Chr.9: 95101582-95101582 on Build GRCh38
rs730881729 Chr.9: 95101741-95101741 on Build GRCh38
rs760413505 Chr.9: 95101693-95101693 on Build GRCh38
rs761796772 Chr.9: 95101680-95101680 on Build GRCh38
rs369636116 Chr.9: 95101720-95101720 on Build GRCh38
rs775430136 Chr.9: 95101774-95101774 on Build GRCh38
rs920493853 Chr.9: 95101602-95101602 on Build GRCh38
rs76575945 Chr.9: 95101669-95101669 on Build GRCh38
rs758866109 Chr.9: 95101712-95101712 on Build GRCh38
rs370974124 Chr.9: 95101721-95101721 on Build GRCh38
rs369235858 Chr.9: 95101637-95101637 on Build GRCh38
rs558379284 Chr.9: 95101567-95101567 on Build GRCh38
rs76092534 Chr.9: 95101670-95101670 on Build GRCh38
rs753428914 Chr.9: 95101699-95101699 on Build GRCh38
rs369503655 Chr.9: 95101661-95101661 on Build GRCh38
rs34831009 Chr.9: 95101670-95101671 on Build GRCh38
rs372511678 Chr.9: 95101700-95101700 on Build GRCh38
rs375362894 Chr.9: 95101761-95101761 on Build GRCh38
rs766410371 Chr.9: 95101762-95101762 on Build GRCh38
rs767209716 Chr.9: 95101691-95101691 on Build GRCh38
rs571668582 Chr.9: 95101750-95101750 on Build GRCh38
rs769743634 Chr.9: 95101724-95101724 on Build GRCh38
rs863224610 Chr.9: 95101715-95101715 on Build GRCh38
rs1015262677 Chr.9: 95101610-95101610 on Build GRCh38
rs934624909 Chr.9: 95101640-95101640 on Build GRCh38
rs1057515705 Chr.9: 95101609-95101610 on Build GRCh38
rs7029888 Chr.9: 95101665-95101665 on Build GRCh38
rs370270817 Chr.9: 95101698-95101698 on Build GRCh38
rs540891064 Chr.9: 95101603-95101603 on Build GRCh38
rs888378164 Chr.9: 95101687-95101687 on Build GRCh38
rs104886458 Chr.9: 95101723-95101723 on Build GRCh38
rs779450392 Chr.9: 95101624-95101624 on Build GRCh38
rs763914550 Chr.9: 95101662-95101662 on Build GRCh38
rs928066874 Chr.9: 95101652-95101652 on Build GRCh38
rs770935196 Chr.9: 95101681-95101681 on Build GRCh38
rs759413874 Chr.9: 95101690-95101690 on Build GRCh38
rs780384904 Chr.9: 95101713-95101713 on Build GRCh38
rs760573194 Chr.9: 95101626-95101626 on Build GRCh38
rs56383909 Chr.9: 95101630-95101630 on Build GRCh38
rs995841421 Chr.9: 95101770-95101770 on Build GRCh38
rs55687573 Chr.9: 95101611-95101611 on Build GRCh38

View Full Product Details