Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150330467-150330740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC61
Entrez Gene ID
65999
Gene Name
leucine rich repeat containing 61
Gene Aliases
HSPC295
UniGene
Hs.647119
Gene Chromosome Location
Chr.7: 150322639-150338156 on Build GRCh38
Transcripts
NM_001142928.1 NM_023942.2 XM_006716095.2 NM_138434.2

Gene Symbol
ZBED6CL
Entrez Gene ID
113763
Gene Name
ZBED6 C-terminal like
Gene Aliases
C7orf29
UniGene
Hs.655915
Gene Chromosome Location
Chr.7: 150329849-150332721 on Build GRCh38
Transcripts
NM_001142928.1 NM_023942.2 XM_006716095.2 NM_138434.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
73964 c.310G>C p.V104L Chr.7: 150330714-150330714 on Build GRCh38
339522 c.123G>C p.E41D Chr.7: 150330527-150330527 on Build GRCh38
7352869 c.85C>T p.Q29* Chr.7: 150330489-150330489 on Build GRCh38
5358145 c.143T>C p.L48P Chr.7: 150330547-150330547 on Build GRCh38
7352870 c.278C>T p.P93L Chr.7: 150330682-150330682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201506127 Chr.7: 150330644-150330644 on Build GRCh38
rs759997199 Chr.7: 150330503-150330503 on Build GRCh38
rs750101404 Chr.7: 150330699-150330699 on Build GRCh38
rs945464067 Chr.7: 150330500-150330500 on Build GRCh38
rs756908891 Chr.7: 150330607-150330607 on Build GRCh38
rs749242548 Chr.7: 150330638-150330638 on Build GRCh38
rs369447873 Chr.7: 150330522-150330522 on Build GRCh38
rs763728477 Chr.7: 150330605-150330605 on Build GRCh38
rs754866897 Chr.7: 150330639-150330639 on Build GRCh38
rs773438604 Chr.7: 150330488-150330488 on Build GRCh38
rs753595180 Chr.7: 150330710-150330710 on Build GRCh38
rs372975857 Chr.7: 150330517-150330517 on Build GRCh38
rs374205555 Chr.7: 150330714-150330714 on Build GRCh38
rs565899229 Chr.7: 150330691-150330691 on Build GRCh38
rs947104618 Chr.7: 150330648-150330648 on Build GRCh38
rs151278452 Chr.7: 150330603-150330603 on Build GRCh38
rs771128702 Chr.7: 150330655-150330655 on Build GRCh38
rs549907418 Chr.7: 150330609-150330609 on Build GRCh38
rs755670047 Chr.7: 150330701-150330701 on Build GRCh38
rs757989158 Chr.7: 150330518-150330518 on Build GRCh38
rs756948650 Chr.7: 150330514-150330515 on Build GRCh38
rs779763201 Chr.7: 150330612-150330612 on Build GRCh38
rs765766782 Chr.7: 150330515-150330515 on Build GRCh38
rs371078860 Chr.7: 150330565-150330565 on Build GRCh38
rs766868694 Chr.7: 150330698-150330698 on Build GRCh38
rs771166097 Chr.7: 150330569-150330569 on Build GRCh38
rs749154624 Chr.7: 150330552-150330552 on Build GRCh38
rs766189677 Chr.7: 150330703-150330703 on Build GRCh38
rs776921441 Chr.7: 150330578-150330578 on Build GRCh38
rs772150075 Chr.7: 150330556-150330556 on Build GRCh38
rs73474332 Chr.7: 150330489-150330489 on Build GRCh38
rs769676026 Chr.7: 150330546-150330546 on Build GRCh38
rs3800782 Chr.7: 150330674-150330674 on Build GRCh38
rs146899479 Chr.7: 150330579-150330579 on Build GRCh38
rs751082684 Chr.7: 150330606-150330606 on Build GRCh38
rs761379985 Chr.7: 150330697-150330697 on Build GRCh38
rs756893925 Chr.7: 150330523-150330523 on Build GRCh38
rs764675338 Chr.7: 150330687-150330687 on Build GRCh38
rs149586894 Chr.7: 150330683-150330683 on Build GRCh38
rs139310721 Chr.7: 150330583-150330583 on Build GRCh38
rs780034010 Chr.7: 150330549-150330549 on Build GRCh38
rs754204725 Chr.7: 150330499-150330499 on Build GRCh38
rs745653603 Chr.7: 150330535-150330535 on Build GRCh38
rs75555338 Chr.7: 150330713-150330713 on Build GRCh38
rs751320383 Chr.7: 150330501-150330506 on Build GRCh38
rs185618761 Chr.7: 150330608-150330608 on Build GRCh38
rs766689528 Chr.7: 150330492-150330492 on Build GRCh38
rs780888874 Chr.7: 150330533-150330533 on Build GRCh38
rs202189755 Chr.7: 150330686-150330686 on Build GRCh38
rs143446716 Chr.7: 150330564-150330564 on Build GRCh38
rs763506194 Chr.7: 150330604-150330604 on Build GRCh38
rs376158905 Chr.7: 150330521-150330521 on Build GRCh38
rs79275116 Chr.7: 150330682-150330682 on Build GRCh38

View Full Product Details