Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41534539-41534782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASK
Entrez Gene ID
8573
Gene Name
calcium/calmodulin dependent serine protein kinase
Gene Aliases
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8
UniGene
Hs.495984
Gene Chromosome Location
Chr.X: 41514934-41923525 on Build GRCh38
Transcripts
NM_001126054.2 NM_001126055.2 NM_003688.3 XM_005272686.4 XM_006724566.3 XM_011543993.2 XM_011543994.2 XM_011543995.2 XM_011543996.2 XM_011543997.2

SNP Detail

SNP ID Chromosome Location
rs754190870 Chr.X: 41534582-41534582 on Build GRCh38
rs758345399 Chr.X: 41534746-41534746 on Build GRCh38
rs754678428 Chr.X: 41534719-41534719 on Build GRCh38
rs797045432 Chr.X: 41534696-41534696 on Build GRCh38
rs146148806 Chr.X: 41534602-41534602 on Build GRCh38
rs137964936 Chr.X: 41534726-41534726 on Build GRCh38
rs374791267 Chr.X: 41534691-41534691 on Build GRCh38
rs765331588 Chr.X: 41534677-41534677 on Build GRCh38
rs373953251 Chr.X: 41534727-41534727 on Build GRCh38
rs933546465 Chr.X: 41534748-41534748 on Build GRCh38
rs398123811 Chr.X: 41534706-41534706 on Build GRCh38
rs749939463 Chr.X: 41534686-41534686 on Build GRCh38
rs746390137 Chr.X: 41534735-41534735 on Build GRCh38
rs765916304 Chr.X: 41534692-41534692 on Build GRCh38
rs886041645 Chr.X: 41534752-41534753 on Build GRCh38
rs5964007 Chr.X: 41534697-41534697 on Build GRCh38
rs906248025 Chr.X: 41534634-41534634 on Build GRCh38
rs747151105 Chr.X: 41534750-41534750 on Build GRCh38

View Full Product Details