Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6708798-6709067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT5
Entrez Gene ID
55326
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 5
Gene Aliases
1AGPAT5, LPAATE
UniGene
Hs.624002
Gene Chromosome Location
Chr.8: 6708357-6761503 on Build GRCh38
Transcripts
NM_018361.3 XM_011534751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755769 c.162C>G p.L54L Chr.8: 6708830-6708830 on Build GRCh38
6515941 c.134G>A p.R45H Chr.8: 6708802-6708802 on Build GRCh38
2720625 c.206A>G p.Y69C Chr.8: 6708874-6708874 on Build GRCh38
297548 c.204T>G p.N68K Chr.8: 6708872-6708872 on Build GRCh38
5358583 c.188_190delTCT p.F66delF Chr.8: 6708856-6708858 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746843517 Chr.8: 6708993-6708993 on Build GRCh38
rs1004440942 Chr.8: 6709026-6709026 on Build GRCh38
rs758996396 Chr.8: 6708849-6708849 on Build GRCh38
rs755286180 Chr.8: 6708912-6708912 on Build GRCh38
rs62497278 Chr.8: 6708830-6708830 on Build GRCh38
rs575564844 Chr.8: 6709030-6709030 on Build GRCh38
rs1015013189 Chr.8: 6709031-6709031 on Build GRCh38
rs769797993 Chr.8: 6708825-6708825 on Build GRCh38
rs370009862 Chr.8: 6708909-6708909 on Build GRCh38
rs374945752 Chr.8: 6708883-6708883 on Build GRCh38
rs752943962 Chr.8: 6708926-6708927 on Build GRCh38
rs779047605 Chr.8: 6708920-6708920 on Build GRCh38
rs757506228 Chr.8: 6708875-6708875 on Build GRCh38
rs531899292 Chr.8: 6708936-6708936 on Build GRCh38
rs758376470 Chr.8: 6708989-6708989 on Build GRCh38
rs764443317 Chr.8: 6708957-6708957 on Build GRCh38
rs536975645 Chr.8: 6708905-6708905 on Build GRCh38
rs770433497 Chr.8: 6708996-6708996 on Build GRCh38
rs376998278 Chr.8: 6708903-6708903 on Build GRCh38
rs764684827 Chr.8: 6708917-6708917 on Build GRCh38
rs1045169178 Chr.8: 6708985-6708985 on Build GRCh38
rs747867622 Chr.8: 6708942-6708942 on Build GRCh38
rs749412458 Chr.8: 6708938-6708938 on Build GRCh38
rs759102557 Chr.8: 6708954-6708954 on Build GRCh38
rs777027450 Chr.8: 6709050-6709050 on Build GRCh38
rs765787531 Chr.8: 6708910-6708910 on Build GRCh38
rs946790542 Chr.8: 6708984-6708984 on Build GRCh38
rs774992583 Chr.8: 6708959-6708959 on Build GRCh38
rs967209405 Chr.8: 6709036-6709036 on Build GRCh38
rs745575559 Chr.8: 6709017-6709017 on Build GRCh38
rs777969983 Chr.8: 6708851-6708851 on Build GRCh38
rs939490116 Chr.8: 6709003-6709003 on Build GRCh38
rs372618285 Chr.8: 6708920-6708920 on Build GRCh38
rs752004741 Chr.8: 6708873-6708873 on Build GRCh38
rs773041597 Chr.8: 6708944-6708944 on Build GRCh38
rs561653572 Chr.8: 6708835-6708835 on Build GRCh38
rs760187760 Chr.8: 6708847-6708847 on Build GRCh38
rs766642940 Chr.8: 6708979-6708979 on Build GRCh38
rs775186694 Chr.8: 6709039-6709039 on Build GRCh38
rs779026470 Chr.8: 6708982-6708982 on Build GRCh38
rs780235451 Chr.8: 6708880-6708880 on Build GRCh38
rs898792965 Chr.8: 6709040-6709040 on Build GRCh38
rs750857299 Chr.8: 6708966-6708966 on Build GRCh38
rs775955230 Chr.8: 6708947-6708947 on Build GRCh38
rs775293063 Chr.8: 6708826-6708826 on Build GRCh38
rs769429848 Chr.8: 6709009-6709009 on Build GRCh38
rs1052554515 Chr.8: 6708822-6708822 on Build GRCh38
rs756247297 Chr.8: 6708971-6708971 on Build GRCh38
rs925939769 Chr.8: 6708931-6708931 on Build GRCh38
rs545501024 Chr.8: 6708980-6708980 on Build GRCh38
rs750658975 Chr.8: 6708933-6708933 on Build GRCh38
rs367932714 Chr.8: 6708878-6708878 on Build GRCh38
rs777170599 Chr.8: 6709045-6709045 on Build GRCh38
rs138274785 Chr.8: 6708839-6708839 on Build GRCh38
rs192900225 Chr.8: 6708928-6708928 on Build GRCh38
rs935238925 Chr.8: 6708958-6708958 on Build GRCh38
rs953174428 Chr.8: 6708918-6708918 on Build GRCh38
rs374017997 Chr.8: 6708848-6708848 on Build GRCh38
rs777508811 Chr.8: 6708992-6708992 on Build GRCh38
rs967431468 Chr.8: 6708923-6708923 on Build GRCh38
rs771843698 Chr.8: 6708943-6708943 on Build GRCh38
rs748058111 Chr.8: 6708986-6708986 on Build GRCh38
rs763014813 Chr.8: 6708827-6708827 on Build GRCh38
rs762378350 Chr.8: 6709047-6709047 on Build GRCh38
rs557194721 Chr.8: 6708915-6708915 on Build GRCh38
rs769758252 Chr.8: 6708879-6708879 on Build GRCh38
rs373598615 Chr.8: 6708901-6708901 on Build GRCh38
rs369624223 Chr.8: 6708893-6708893 on Build GRCh38
rs753432175 Chr.8: 6708914-6708914 on Build GRCh38
rs767971723 Chr.8: 6708962-6708962 on Build GRCh38
rs1046120435 Chr.8: 6708836-6708836 on Build GRCh38
rs778832590 Chr.8: 6708939-6708939 on Build GRCh38
rs776352644 Chr.8: 6709008-6709008 on Build GRCh38
rs565304170 Chr.8: 6709012-6709012 on Build GRCh38
rs1028898072 Chr.8: 6708998-6708998 on Build GRCh38
rs887377403 Chr.8: 6709018-6709018 on Build GRCh38
rs754210181 Chr.8: 6708856-6708858 on Build GRCh38

View Full Product Details