Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 69505833-69506068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM155B
Entrez Gene ID
27112
Gene Name
family with sequence similarity 155 member B
Gene Aliases
CXorf63, TED, TMEM28, bB57D9.1
UniGene
Hs.87619
Gene Chromosome Location
Chr.X: 69505204-69532508 on Build GRCh38
Transcripts
NM_015686.2 XM_011530908.2 XM_011530909.2

SNP Detail

SNP ID Chromosome Location
rs146856746 Chr.X: 69505973-69505973 on Build GRCh38
rs772582182 Chr.X: 69505995-69505995 on Build GRCh38
rs374491730 Chr.X: 69505942-69505942 on Build GRCh38
rs917671957 Chr.X: 69506026-69506026 on Build GRCh38
rs140715552 Chr.X: 69505981-69505981 on Build GRCh38
rs763953914 Chr.X: 69506039-69506039 on Build GRCh38
rs141000146 Chr.X: 69505952-69505952 on Build GRCh38
rs776253229 Chr.X: 69505955-69505955 on Build GRCh38
rs780282770 Chr.X: 69505939-69505939 on Build GRCh38
rs756307961 Chr.X: 69505909-69505909 on Build GRCh38
rs758991107 Chr.X: 69505957-69505957 on Build GRCh38
rs201151491 Chr.X: 69505943-69505943 on Build GRCh38
rs371709320 Chr.X: 69506028-69506028 on Build GRCh38
rs201772031 Chr.X: 69505997-69505997 on Build GRCh38
rs755330269 Chr.X: 69505993-69505993 on Build GRCh38
rs759273141 Chr.X: 69505856-69505856 on Build GRCh38
rs867076028 Chr.X: 69505911-69505911 on Build GRCh38
rs769444908 Chr.X: 69506016-69506016 on Build GRCh38
rs200299714 Chr.X: 69505994-69505994 on Build GRCh38
rs752493079 Chr.X: 69505866-69505866 on Build GRCh38
rs756149484 Chr.X: 69505978-69505978 on Build GRCh38
rs1005425404 Chr.X: 69506017-69506017 on Build GRCh38
rs748720224 Chr.X: 69505951-69505951 on Build GRCh38
rs371432784 Chr.X: 69505894-69505894 on Build GRCh38
rs764041624 Chr.X: 69505970-69505970 on Build GRCh38
rs995425500 Chr.X: 69505870-69505870 on Build GRCh38
rs758302010 Chr.X: 69505880-69505880 on Build GRCh38
rs773625287 Chr.X: 69505996-69505996 on Build GRCh38
rs199640674 Chr.X: 69505924-69505924 on Build GRCh38
rs759359643 Chr.X: 69505975-69505975 on Build GRCh38
rs905115266 Chr.X: 69505867-69505867 on Build GRCh38
rs767373488 Chr.X: 69505974-69505974 on Build GRCh38
rs775289057 Chr.X: 69505961-69505961 on Build GRCh38
rs774234254 Chr.X: 69506043-69506043 on Build GRCh38
rs749585287 Chr.X: 69505941-69505941 on Build GRCh38
rs149821108 Chr.X: 69505958-69505958 on Build GRCh38
rs751542420 Chr.X: 69505972-69505972 on Build GRCh38
rs764919201 Chr.X: 69505864-69505864 on Build GRCh38
rs368057024 Chr.X: 69505963-69505963 on Build GRCh38
rs1055959173 Chr.X: 69505953-69505953 on Build GRCh38
rs190801973 Chr.X: 69505987-69505987 on Build GRCh38
rs764059847 Chr.X: 69505885-69505885 on Build GRCh38
rs151056099 Chr.X: 69505904-69505904 on Build GRCh38
rs775270963 Chr.X: 69506023-69506023 on Build GRCh38

View Full Product Details