Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113800672-113800907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM255B
Entrez Gene ID
348013
Gene Name
transmembrane protein 255B
Gene Aliases
FAM70B
UniGene
Hs.280805
Gene Chromosome Location
Chr.13: 113759217-113813797 on Build GRCh38
Transcripts
NM_182614.2 XM_005272413.3 XM_005272415.2 XM_006719986.2 XM_017020558.1 XM_017020559.1 XM_017020560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6414470 c.493C>T p.L165F Chr.13: 113800896-113800896 on Build GRCh38
3467444 c.464T>G p.V155G Chr.13: 113800867-113800867 on Build GRCh38
6531826 c.484T>C p.C162R Chr.13: 113800887-113800887 on Build GRCh38
3417398 c.490G>T p.D164Y Chr.13: 113800893-113800893 on Build GRCh38
5376458 c.459G>A p.L153L Chr.13: 113800862-113800862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985722336 Chr.13: 113800700-113800700 on Build GRCh38
rs573922382 Chr.13: 113800694-113800694 on Build GRCh38
rs778414031 Chr.13: 113800784-113800784 on Build GRCh38
rs545611614 Chr.13: 113800769-113800769 on Build GRCh38
rs201537526 Chr.13: 113800847-113800847 on Build GRCh38
rs763522174 Chr.13: 113800821-113800821 on Build GRCh38
rs1008588750 Chr.13: 113800811-113800811 on Build GRCh38
rs541398344 Chr.13: 113800778-113800779 on Build GRCh38
rs374555220 Chr.13: 113800885-113800885 on Build GRCh38
rs1018513456 Chr.13: 113800708-113800708 on Build GRCh38
rs780977012 Chr.13: 113800831-113800831 on Build GRCh38
rs907970535 Chr.13: 113800702-113800702 on Build GRCh38
rs774585714 Chr.13: 113800875-113800875 on Build GRCh38
rs899047642 Chr.13: 113800762-113800762 on Build GRCh38
rs757868406 Chr.13: 113800793-113800793 on Build GRCh38
rs771084796 Chr.13: 113800860-113800860 on Build GRCh38
rs756567658 Chr.13: 113800776-113800776 on Build GRCh38
rs372861793 Chr.13: 113800798-113800798 on Build GRCh38
rs113798460 Chr.13: 113800699-113800699 on Build GRCh38
rs958224089 Chr.13: 113800868-113800868 on Build GRCh38
rs578125837 Chr.13: 113800734-113800734 on Build GRCh38
rs1021823806 Chr.13: 113800693-113800693 on Build GRCh38
rs940489814 Chr.13: 113800735-113800735 on Build GRCh38
rs58284072 Chr.13: 113800794-113800794 on Build GRCh38
rs965374095 Chr.13: 113800773-113800773 on Build GRCh38
rs377104174 Chr.13: 113800789-113800789 on Build GRCh38
rs559546736 Chr.13: 113800718-113800718 on Build GRCh38
rs147264445 Chr.13: 113800848-113800848 on Build GRCh38
rs760051716 Chr.13: 113800879-113800879 on Build GRCh38
rs1037505056 Chr.13: 113800759-113800759 on Build GRCh38
rs541529601 Chr.13: 113800697-113800697 on Build GRCh38
rs189764657 Chr.13: 113800842-113800842 on Build GRCh38

View Full Product Details