Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87971136-87971383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK20
Entrez Gene ID
23552
Gene Name
cyclin dependent kinase 20
Gene Aliases
CCRK, CDCH, P42, PNQALRE
UniGene
Hs.522274
Gene Chromosome Location
Chr.9: 87966441-87974780 on Build GRCh38
Transcripts
NM_001039803.2 NM_001170639.1 NM_001170640.1 NM_012119.4 NM_178432.3 XM_017014561.1 XM_017014562.1 XM_017014563.1 XM_017014564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3659379 c.378G>A p.R126R Chr.9: 87971147-87971147 on Build GRCh38
3659384 c.329G>A p.R110K Chr.9: 87971235-87971235 on Build GRCh38
3659383 c.290G>A p.R97K Chr.9: 87971235-87971235 on Build GRCh38
3659381 c.345C>T p.V115V Chr.9: 87971180-87971180 on Build GRCh38
5876536 c.297_298GG>AA p.D100N Chr.9: 87971266-87971267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748362044 Chr.9: 87971332-87971332 on Build GRCh38
rs775356611 Chr.9: 87971298-87971298 on Build GRCh38
rs564653137 Chr.9: 87971254-87971254 on Build GRCh38
rs768885451 Chr.9: 87971241-87971241 on Build GRCh38
rs746419285 Chr.9: 87971231-87971231 on Build GRCh38
rs772151523 Chr.9: 87971293-87971293 on Build GRCh38
rs775554830 Chr.9: 87971345-87971345 on Build GRCh38
rs760742841 Chr.9: 87971355-87971355 on Build GRCh38
rs777370245 Chr.9: 87971190-87971190 on Build GRCh38
rs3750491 Chr.9: 87971240-87971240 on Build GRCh38
rs772400848 Chr.9: 87971232-87971232 on Build GRCh38
rs755864917 Chr.9: 87971186-87971186 on Build GRCh38
rs766346750 Chr.9: 87971164-87971164 on Build GRCh38
rs774327447 Chr.9: 87971252-87971252 on Build GRCh38
rs759499348 Chr.9: 87971292-87971292 on Build GRCh38
rs757636291 Chr.9: 87971202-87971202 on Build GRCh38
rs373884873 Chr.9: 87971276-87971276 on Build GRCh38
rs762743425 Chr.9: 87971160-87971160 on Build GRCh38
rs763841221 Chr.9: 87971260-87971260 on Build GRCh38
rs750242727 Chr.9: 87971281-87971282 on Build GRCh38
rs746350961 Chr.9: 87971271-87971271 on Build GRCh38
rs777853462 Chr.9: 87971336-87971336 on Build GRCh38
rs1015277903 Chr.9: 87971325-87971325 on Build GRCh38
rs892523635 Chr.9: 87971215-87971215 on Build GRCh38
rs139270796 Chr.9: 87971267-87971267 on Build GRCh38
rs776720051 Chr.9: 87971159-87971159 on Build GRCh38
rs144310084 Chr.9: 87971247-87971247 on Build GRCh38
rs370419416 Chr.9: 87971344-87971344 on Build GRCh38
rs758842628 Chr.9: 87971312-87971312 on Build GRCh38
rs781454036 Chr.9: 87971331-87971331 on Build GRCh38
rs9410653 Chr.9: 87971306-87971306 on Build GRCh38
rs752120731 Chr.9: 87971317-87971317 on Build GRCh38
rs369580008 Chr.9: 87971225-87971225 on Build GRCh38
rs28364953 Chr.9: 87971268-87971268 on Build GRCh38
rs761951896 Chr.9: 87971248-87971248 on Build GRCh38
rs146852927 Chr.9: 87971288-87971288 on Build GRCh38
rs755317703 Chr.9: 87971320-87971320 on Build GRCh38
rs767199625 Chr.9: 87971266-87971266 on Build GRCh38
rs373501905 Chr.9: 87971234-87971234 on Build GRCh38
rs938184258 Chr.9: 87971284-87971284 on Build GRCh38
rs143756400 Chr.9: 87971275-87971275 on Build GRCh38
rs775868376 Chr.9: 87971239-87971239 on Build GRCh38
rs749920703 Chr.9: 87971308-87971308 on Build GRCh38
rs918242543 Chr.9: 87971249-87971249 on Build GRCh38
rs377363483 Chr.9: 87971343-87971343 on Build GRCh38
rs752366494 Chr.9: 87971182-87971182 on Build GRCh38
rs200828251 Chr.9: 87971251-87971251 on Build GRCh38
rs781467120 Chr.9: 87971278-87971278 on Build GRCh38
rs769886503 Chr.9: 87971290-87971290 on Build GRCh38
rs945495949 Chr.9: 87971243-87971243 on Build GRCh38
rs761408768 Chr.9: 87971305-87971305 on Build GRCh38
rs760540412 Chr.9: 87971302-87971302 on Build GRCh38
rs376766367 Chr.9: 87971259-87971259 on Build GRCh38
rs376523146 Chr.9: 87971180-87971180 on Build GRCh38
rs769932384 Chr.9: 87971335-87971335 on Build GRCh38
rs759432942 Chr.9: 87971257-87971257 on Build GRCh38
rs780536384 Chr.9: 87971315-87971315 on Build GRCh38
rs896746701 Chr.9: 87971296-87971296 on Build GRCh38
rs758932557 Chr.9: 87971274-87971274 on Build GRCh38
rs199935579 Chr.9: 87971258-87971258 on Build GRCh38
rs748842542 Chr.9: 87971195-87971195 on Build GRCh38
rs41286029 Chr.9: 87971208-87971208 on Build GRCh38
rs965150299 Chr.9: 87971197-87971197 on Build GRCh38
rs764037601 Chr.9: 87971304-87971304 on Build GRCh38
rs200737032 Chr.9: 87971209-87971209 on Build GRCh38
rs140703002 Chr.9: 87971291-87971291 on Build GRCh38
rs973836002 Chr.9: 87971334-87971334 on Build GRCh38
rs751521835 Chr.9: 87971171-87971171 on Build GRCh38
rs754792892 Chr.9: 87971179-87971179 on Build GRCh38

View Full Product Details