Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98128087-98128340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO2A
Entrez Gene ID
7464
Gene Name
coronin 2A
Gene Aliases
CLIPINB, IR10, WDR2
UniGene
Hs.113094
Gene Chromosome Location
Chr.9: 98120975-98192674 on Build GRCh38
Transcripts
NM_003389.3 NM_052820.3 XM_011518986.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3925758 c.1149G>A p.E383E Chr.9: 98128192-98128192 on Build GRCh38
3652195 c.1130C>T p.P377L Chr.9: 98128211-98128211 on Build GRCh38
6321552 c.1129C>A p.P377T Chr.9: 98128212-98128212 on Build GRCh38
455024 c.1096G>A p.E366K Chr.9: 98128245-98128245 on Build GRCh38
5410016 c.1140G>C p.T380T Chr.9: 98128201-98128201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778191201 Chr.9: 98128298-98128298 on Build GRCh38
rs755199037 Chr.9: 98128263-98128263 on Build GRCh38
rs34014018 Chr.9: 98128113-98128113 on Build GRCh38
rs201111862 Chr.9: 98128172-98128172 on Build GRCh38
rs770298492 Chr.9: 98128186-98128186 on Build GRCh38
rs753085117 Chr.9: 98128290-98128290 on Build GRCh38
rs780182594 Chr.9: 98128175-98128175 on Build GRCh38
rs150107665 Chr.9: 98128288-98128288 on Build GRCh38
rs770513543 Chr.9: 98128130-98128130 on Build GRCh38
rs771924987 Chr.9: 98128206-98128206 on Build GRCh38
rs750489715 Chr.9: 98128171-98128171 on Build GRCh38
rs759401045 Chr.9: 98128204-98128204 on Build GRCh38
rs755397566 Chr.9: 98128179-98128179 on Build GRCh38
rs770593745 Chr.9: 98128201-98128201 on Build GRCh38
rs766614555 Chr.9: 98128257-98128257 on Build GRCh38
rs756705426 Chr.9: 98128240-98128240 on Build GRCh38
rs752748473 Chr.9: 98128160-98128160 on Build GRCh38
rs12375882 Chr.9: 98128173-98128173 on Build GRCh38
rs762995358 Chr.9: 98128254-98128254 on Build GRCh38
rs781664363 Chr.9: 98128182-98128182 on Build GRCh38
rs748781900 Chr.9: 98128198-98128198 on Build GRCh38
rs777290329 Chr.9: 98128193-98128193 on Build GRCh38
rs748872959 Chr.9: 98128316-98128316 on Build GRCh38
rs1056183650 Chr.9: 98128304-98128304 on Build GRCh38
rs751850075 Chr.9: 98128261-98128261 on Build GRCh38
rs775434600 Chr.9: 98128233-98128233 on Build GRCh38
rs907792053 Chr.9: 98128180-98128180 on Build GRCh38
rs753793631 Chr.9: 98128297-98128297 on Build GRCh38
rs774164287 Chr.9: 98128136-98128136 on Build GRCh38
rs373658313 Chr.9: 98128122-98128122 on Build GRCh38
rs760636423 Chr.9: 98128235-98128235 on Build GRCh38
rs541402540 Chr.9: 98128113-98128113 on Build GRCh38
rs763967937 Chr.9: 98128295-98128296 on Build GRCh38
rs11794723 Chr.9: 98128309-98128309 on Build GRCh38
rs77416773 Chr.9: 98128202-98128202 on Build GRCh38
rs143294637 Chr.9: 98128183-98128183 on Build GRCh38
rs1019026501 Chr.9: 98128116-98128116 on Build GRCh38
rs758476447 Chr.9: 98128285-98128285 on Build GRCh38
rs2231668 Chr.9: 98128121-98128121 on Build GRCh38
rs751932580 Chr.9: 98128178-98128178 on Build GRCh38
rs981515747 Chr.9: 98128234-98128234 on Build GRCh38
rs751448430 Chr.9: 98128299-98128300 on Build GRCh38
rs756784251 Chr.9: 98128307-98128307 on Build GRCh38
rs574027996 Chr.9: 98128109-98128109 on Build GRCh38
rs759452151 Chr.9: 98128139-98128139 on Build GRCh38
rs760690885 Chr.9: 98128167-98128167 on Build GRCh38
rs773309381 Chr.9: 98128242-98128242 on Build GRCh38
rs765299966 Chr.9: 98128169-98128169 on Build GRCh38
rs748104776 Chr.9: 98128236-98128236 on Build GRCh38
rs767548306 Chr.9: 98128151-98128151 on Build GRCh38
rs748815887 Chr.9: 98128302-98128302 on Build GRCh38

View Full Product Details