Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99127080-99127341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH4
Entrez Gene ID
127
Gene Name
alcohol dehydrogenase 4 (class II), pi polypeptide
Gene Aliases
ADH-2, HEL-S-4
UniGene
Hs.1219
Gene Chromosome Location
Chr.4: 99123657-99144330 on Build GRCh38
Transcripts
NM_000670.4 NM_001306171.1 NM_001306172.1 XM_017007713.1 XM_017007714.1 XM_017007715.1 NR_037884.1

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NM_000670.4 NM_001306171.1 NM_001306172.1 XM_017007713.1 XM_017007714.1 XM_017007715.1 NR_037884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7106574 c.991G>A p.E331K Chr.4: 99127254-99127254 on Build GRCh38
5680780 c.1033G>A p.G345S Chr.4: 99127212-99127212 on Build GRCh38
1182069 c.970T>A p.F324I Chr.4: 99127218-99127218 on Build GRCh38
6331948 c.932G>T p.W311L Chr.4: 99127313-99127313 on Build GRCh38
6165988 c.990A>G p.P330P Chr.4: 99127255-99127255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4148891 Chr.4: 99127194-99127194 on Build GRCh38
rs772707994 Chr.4: 99127160-99127160 on Build GRCh38
rs764200531 Chr.4: 99127290-99127290 on Build GRCh38
rs746932989 Chr.4: 99127176-99127176 on Build GRCh38
rs777089873 Chr.4: 99127260-99127260 on Build GRCh38
rs927992570 Chr.4: 99127279-99127279 on Build GRCh38
rs758416313 Chr.4: 99127204-99127204 on Build GRCh38
rs9307234 Chr.4: 99127176-99127176 on Build GRCh38
rs765340669 Chr.4: 99127187-99127187 on Build GRCh38
rs773792758 Chr.4: 99127167-99127167 on Build GRCh38
rs755804627 Chr.4: 99127315-99127315 on Build GRCh38
rs200898633 Chr.4: 99127262-99127262 on Build GRCh38
rs779502378 Chr.4: 99127243-99127243 on Build GRCh38
rs750236433 Chr.4: 99127312-99127312 on Build GRCh38
rs370189152 Chr.4: 99127169-99127169 on Build GRCh38
rs754657743 Chr.4: 99127322-99127322 on Build GRCh38
rs554366907 Chr.4: 99127203-99127203 on Build GRCh38
rs764147461 Chr.4: 99127221-99127221 on Build GRCh38
rs147130103 Chr.4: 99127232-99127232 on Build GRCh38
rs776287384 Chr.4: 99127193-99127194 on Build GRCh38
rs928612218 Chr.4: 99127290-99127292 on Build GRCh38
rs957983649 Chr.4: 99127210-99127210 on Build GRCh38
rs920591379 Chr.4: 99127296-99127296 on Build GRCh38
rs1028205466 Chr.4: 99127189-99127189 on Build GRCh38
rs771296326 Chr.4: 99127259-99127259 on Build GRCh38
rs200208857 Chr.4: 99127320-99127320 on Build GRCh38
rs371092789 Chr.4: 99127186-99127186 on Build GRCh38
rs557396464 Chr.4: 99127135-99127135 on Build GRCh38
rs767318009 Chr.4: 99127306-99127306 on Build GRCh38
rs371775374 Chr.4: 99127321-99127321 on Build GRCh38
rs766810583 Chr.4: 99127173-99127173 on Build GRCh38
rs938001268 Chr.4: 99127292-99127292 on Build GRCh38
rs192270953 Chr.4: 99127240-99127240 on Build GRCh38
rs770070155 Chr.4: 99127267-99127267 on Build GRCh38
rs748815612 Chr.4: 99127244-99127244 on Build GRCh38
rs930904195 Chr.4: 99127302-99127302 on Build GRCh38
rs778559919 Chr.4: 99127256-99127256 on Build GRCh38
rs775747592 Chr.4: 99127269-99127269 on Build GRCh38
rs29001219 Chr.4: 99127235-99127235 on Build GRCh38
rs1126671 Chr.4: 99127263-99127263 on Build GRCh38
rs781087602 Chr.4: 99127236-99127236 on Build GRCh38
rs200069768 Chr.4: 99127305-99127305 on Build GRCh38
rs747570796 Chr.4: 99127258-99127258 on Build GRCh38
rs761406306 Chr.4: 99127124-99127124 on Build GRCh38
rs187103819 Chr.4: 99127130-99127130 on Build GRCh38
rs768337448 Chr.4: 99127251-99127251 on Build GRCh38
rs763160008 Chr.4: 99127281-99127281 on Build GRCh38
rs367861880 Chr.4: 99127295-99127295 on Build GRCh38
rs374404089 Chr.4: 99127239-99127239 on Build GRCh38

View Full Product Details